Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511811_m1
          See other 1700029M20RIK GE Assays ›
          Gene Symbol:
          1700029M20Rik
          Gene Name:
          RIKEN cDNA 1700029M20 gene
          Gene Aliases:
          4930406E24Rik
          Chromosome Location:
          Chr.4: 135626655 - 135630198 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511811_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700029M20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027909.1 1 - 2 390 75
          GenBank mRNA AK015107.1 - 1 - 2 228 75
          BC059720.1 - 1 - 2 249 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA NM_028688.1 - 228 Chr. - - [-] on Build 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001660.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700029M20Rik

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2263342 AK006510.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2263342 NR_015613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2263342 NR_027909.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848120 U55872.1 genbank_mrna_mouse
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2263342 AK006510.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2263342 NR_015613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2263342 NR_027909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208367 AK006510.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208367 AK015107.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208367 AK016438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208367 NR_015613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208367 NR_027909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453456 AK006510.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453456 NR_015613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P453456 NR_027909.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03476797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03554051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03645263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03457203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03554074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03414122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03926323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03554056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03444264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03871119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03645267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03871117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03876439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03404206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23329686 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10509050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509051 - -
          Affymetrix HT MG-430 PM Array Plate 1453975_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1453154_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10509049 - -
          Mouse Genome 430 2.0 Array 1453154_PM_at - -
          Mouse Genome 430 2.0 Array 1453154_at - -
          Mouse Genome 430 2.0 Array 1453975_a_at - -
          Mouse Genome 430 2.0 Array 1453975_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR0400013214.mm.1 NR_015613.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013214.mm.1 NR_015613.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013214.mm.1 NR_027909.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013214.mm.1 NR_027909.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013214.mm.1 TC0400001660.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400013216.mm.1 NR_015613.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013216.mm.1 NR_015613.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013216.mm.1 NR_027909.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013216.mm.1 NR_027909.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013216.mm.1 TC0400001660.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400013215.mm.1 NR_015613.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013215.mm.1 NR_015613.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013215.mm.1 NR_027909.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013215.mm.1 NR_027909.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013215.mm.1 TC0400001660.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400013213.mm.1 NR_015613.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013213.mm.1 NR_015613.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013213.mm.1 NR_027909.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013213.mm.1 NR_027909.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013213.mm.1 TC0400001660.mm.1 MTA TC
          Mouse Expression Set 430 1453975_PM_a_at - -
          Mouse Expression Set 430 1453154_at - -
          Mouse Expression Set 430 1453154_PM_at - -
          Mouse Expression Set 430 1453975_a_at - -
          Murine Genome U74v2 Set 167931_f_at - -
          Murine Genome U74v2 Set 167931_PM_f_at - -
          Murine Genome U74v2 Set 166106_f_at - -
          Murine Genome U74v2 Set 166106_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1453154_at - -
          Affymetrix HT Mouse Genome 430B Array 1453975_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1453975_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1453154_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 725809 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105910278 - -
          Illumina mouse-6 v1.0 expression beadchip 105420072 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2418420 - -
          mouse-6 v1.1 (Illumina) scl0002660.1_97-S - -
          Illumina Sentrix mouse-6.v1 bead array 105420072 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105420072 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline