Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512236_m1
          See other DNAJC14 GE Assays ›
          Gene Symbol:
          Dnajc14
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily C, member 14
          Gene Aliases:
          5730551F12Rik, DNAJ, DRIP78, HDJ3, LIP6
          Chromosome Location:
          Chr.10: 128805468 - 128819446 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512236_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajc14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028873.4 NP_083149.3 2 - 3 1494 109
          GenBank mRNA AK017830.1 - 2 - 3 1508 109
          AK032681.1 - 2 - 3 1476 109
          AK045345.1 - 2 - 3 1551 109
          AK045445.1 - 2 - 3 1588 109
          AK053314.1 - 2 - 3 1556 109
          AK082579.1 - 2 - 3 1495 109
          AK083878.1 - 2 - 3 1506 109
          AK159388.1 - 2 - 3 1507 109
          AK168935.1 - 2 - 3 1586 109
          BC011146.1 - 2 - 3 1559 4016209 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001622.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnajc14

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          protein transport
          G-protein coupled receptor binding
          dopamine receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543296 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543295 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117515 AK082579.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117515 NM_028873.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039896 AK082579.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039896 NM_028873.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 560506 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291517 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635403 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570655 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46598 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157344 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039896 AK082579.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039896 NM_028873.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117515 AK082579.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117515 NM_028873.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185775 BC011146.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543338 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07234162 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07234173 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07234178 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157344 - -
          Whole Mouse Genome G4122A A_51_P185775 BC011146.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95921 AK082579.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95921 AK083878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P95921 NM_028873.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131213 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157344 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08773772 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08773788 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543296 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08295157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08773772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08480429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08480432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08773793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08378480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08773771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08773774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08916725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543328 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10367419 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373579 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367416 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367414 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367415 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367420 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367418 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367421 - -
          Affymetrix HT MG-430 PM Array Plate 1437547_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1437546_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1426553_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10367413 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373577 - -
          Mouse Genome 430 2.0 Array 1437547_PM_s_at - -
          Mouse Genome 430 2.0 Array 1437546_at - -
          Mouse Genome 430 2.0 Array 1426553_PM_at - -
          Mouse Genome 430 2.0 Array 1437547_s_at - -
          Mouse Genome 430 2.0 Array 1426553_at - -
          Mouse Genome 430 2.0 Array 1437546_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74330_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74330_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000011929.mm.1 NM_028873.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011929.mm.1 NM_028873.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011929.mm.1 TC1000001622.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011932.mm.1 NM_028873.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011932.mm.1 NM_028873.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011932.mm.1 TC1000001622.mm.1 MTA TC
          Mouse Expression Set 430 1426553_PM_at - -
          Mouse Expression Set 430 1437547_PM_s_at - -
          Mouse Expression Set 430 1437546_PM_at - -
          Mouse Expression Set 430 1437547_s_at - -
          Mouse Expression Set 430 1437546_at - -
          Mouse Expression Set 430 1426553_at - -
          Mouse Genome 430A 2.0 Array 1437546_PM_at - -
          Mouse Genome 430A 2.0 Array 1437546_at - -
          Mouse Genome 430A 2.0 Array 1437547_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1437547_s_at - -
          Mouse Genome 430A 2.0 Array 1426553_PM_at - -
          Mouse Genome 430A 2.0 Array 1426553_at - -
          Affymetrix HT Mouse Genome 430A Array 1437546_at - -
          Affymetrix HT Mouse Genome 430A Array 1437546_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426553_at - -
          Affymetrix HT Mouse Genome 430A Array 1437547_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1437547_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1426553_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 332625 - -
          ABI Mouse Genome Survey Microarray 908702 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 103780324 - -
          Illumina mouse-6 v1.0 expression beadchip 6420369 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006420369 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2753843 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2753843 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256582 - -
          mouse-6 v1.1 (Illumina) scl38270.7_86-S - -
          Illumina Sentrix mouse-6.v1 bead array 103780324 - -
          Illumina Sentrix mouse-6.v1 bead array 6420369 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38270.7_86-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6420369 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103780324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2753843 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2753843 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2572395 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl38270.7_86-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline