Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512445_m1
          See other OCSTAMP GE Assays ›
          Gene Symbol:
          Ocstamp
          Gene Name:
          osteoclast stimulatory transmembrane protein
          Gene Aliases:
          4833422F24Rik, OC-STAMP
          Chromosome Location:
          Chr.2: 165395450 - 165400394 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512445_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ocstamp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029021.1 NP_083297.1 1 - 2 110 73
          GenBank mRNA AK014751.1 - 1 - 2 110 73
          AK042271.1 - 1 - 2 122 73
          BC020160.1 - 1 - 2 78 3966086 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005149.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ocstamp

          Gene Ontology Categories:

          Function(s) Process(es)

          cell differentiation
          positive regulation of macrophage fusion
          positive regulation of osteoclast differentiation
          cellular response to tumor necrosis factor
          cellular response to estrogen stimulus
          multinuclear osteoclast differentiation
          positive regulation of osteoclast proliferation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21823610 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21823608 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21823609 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P360004 AK032330.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P360004 NM_029021.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P360004 AK014751.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P360004 AK032330.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P360004 NM_029021.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P360004 AK014751.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 957497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 258009 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133278 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P360004 AK032330.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P360004 NM_029021.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P360004 AK014751.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P360004 AK032330.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P360004 NM_029021.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P360004 AK014751.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21823669 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133278 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04774729 - -
          Whole Mouse Genome G4122A A_51_P360004 AK032330.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P360004 NM_029021.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P360004 AK014751.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P360004 AK032330.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P360004 NM_029021.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P360004 AK014751.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499499 AK032330.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499499 AK042271.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499499 BC020160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499499 NM_029021.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499499 AK014751.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105296 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133278 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01954538 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01753661 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01753658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21823608 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21823609 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21823610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21823543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01293035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01293034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01819231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01220648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01919354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02197319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01819234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02155274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21823615 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10489702 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489703 - -
          Affymetrix Mouse Gene 1.0 ST Array 10489704 - -
          Affymetrix HT MG-430 PM Array Plate 1425386_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10489701 - -
          Mouse Genome 430 2.0 Array 1425386_at - -
          Mouse Genome 430 2.0 Array 1425386_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74614_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74614_at - -
          Affymetrix Mouse Clariom S Array PSR0200044001.mm.1 NM_029021.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044001.mm.1 NM_029021.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044001.mm.1 TC0200005149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044003.mm.1 NM_029021.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044003.mm.1 NM_029021.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044003.mm.1 TC0200005149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044002.mm.1 NM_029021.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044002.mm.1 NM_029021.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044002.mm.1 TC0200005149.mm.1 MTA TC
          Mouse Expression Set 430 1425386_at - -
          Mouse Expression Set 430 1425386_PM_at - -
          Mouse Genome 430A 2.0 Array 1425386_PM_at - -
          Mouse Genome 430A 2.0 Array 1425386_at - -
          Affymetrix HT Mouse Genome 430A Array 1425386_at - -
          Affymetrix HT Mouse Genome 430A Array 1425386_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 555119 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4760609 - -
          Illumina mouse-6 v1.0 expression beadchip 106660725 - -
          Illumina mouse-6 v1.0 expression beadchip 4760609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004760609 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2652131 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2493715 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2652131 - -
          mouse-6 v1.1 (Illumina) scl0074614.2_41-S - -
          mouse-6 v1.1 (Illumina) scl18331.3_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 106660725 - -
          Illumina Sentrix mouse-6.v1 bead array 4760609 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0074614.2_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4760609 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106660725 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2493715 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2493715 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline