Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512504_g1
          See other DDIT4 GE Assays ›
          Gene Symbol:
          Ddit4
          Gene Name:
          DNA-damage-inducible transcript 4
          Gene Aliases:
          5830413E08Rik, AA415483, REDD1, Rtp801, dig2
          Chromosome Location:
          Chr.10: 59949675 - 59951770 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512504_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddit4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029083.2 NP_083359.1 2 - 3 326 64
          GenBank mRNA AK017926.1 - 2 - 3 326 64
          AK081046.1 - 2 - 3 390 64
          AY260552.1 - 1 - 2 249 64
          BC131992.1 - 2 - 3 325 40130269 64
          BC132645.1 - 2 - 3 325 40130922 64
          BC145370.1 - 2 - 3 325 9053928 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002224.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddit4

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          neuron migration
          apoptotic process
          brain development
          cell proliferation
          negative regulation of signal transduction
          negative regulation of peptidyl-threonine phosphorylation
          neuron differentiation
          negative regulation of TOR signaling
          negative regulation of peptidyl-serine phosphorylation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          protein complex disassembly
          negative regulation of glycolytic process
          neurotrophin TRK receptor signaling pathway
          defense response to virus
          reactive oxygen species metabolic process
          positive regulation of neuron death
          negative regulation of intracellular signal transduction
          14-3-3 protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171638 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171640 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171634 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171639 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171635 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171642 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171636 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P245796 AK017926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P245796 NM_029083.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P245796 AK017926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P245796 NM_029083.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776496 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136126 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P245796 AK017926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P245796 NM_029083.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P245796 AK017926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P245796 NM_029083.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245796 AK017926.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245796 NM_029083.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245796 AK017926.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245796 NM_029083.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27171639 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136126 - -
          Whole Mouse Genome G4122A A_51_P245796 AK017926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P245796 NM_029083.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P245796 AK017926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P245796 NM_029083.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136322 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136126 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08834127 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08834152 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08834133 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08834118 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08834146 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08834135 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171634 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171639 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171640 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171635 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171642 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171636 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171638 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27171643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27171631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08804445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08322483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08630033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08539050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08834161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08834118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08729813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08480444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08608097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08537378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08729811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08539043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08630043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08458320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27171652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08834127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08834146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08834123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08834156 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10369292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369291 - -
          Affymetrix HT MG-430 PM Array Plate 1428306_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10369290 - -
          Mouse Genome 430 2.0 Array 1428306_at - -
          Mouse Genome 430 2.0 Array 1428306_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74747_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74747_at - -
          Affymetrix Mouse Clariom S Array PSR1000015565.mm.1 NM_029083.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015565.mm.1 NM_029083.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015565.mm.1 TC1000002224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015562.mm.1 NM_029083.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015562.mm.1 NM_029083.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015562.mm.1 TC1000002224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015563.mm.1 NM_029083.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015563.mm.1 NM_029083.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015563.mm.1 TC1000002224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015564.mm.1 NM_029083.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015564.mm.1 NM_029083.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015564.mm.1 TC1000002224.mm.1 MTA TC
          Mouse Expression Set 430 1428306_at - -
          Mouse Expression Set 430 1428306_PM_at - -
          Murine Genome U74v2 Set 103460_at - -
          Murine Genome U74v2 Set 103460_PM_at - -
          Mouse Genome 430A 2.0 Array 1428306_at - -
          Mouse Genome 430A 2.0 Array 1428306_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428306_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428306_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2570408 - -
          Illumina mouse-6 v1.0 expression beadchip 2570408 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570408 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2734764 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2734764 - -
          mouse-6 v1.1 (Illumina) scl37934.3_287-S - -
          Illumina Sentrix mouse-6.v1 bead array 2570408 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37934.3_287-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570408 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2993109 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2993109 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37934.3_287-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline