Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512580_m1
          See other NXNL2 GE Assays ›
          Gene Symbol:
          Nxnl2
          Gene Name:
          nucleoredoxin-like 2
          Gene Aliases:
          4930519N16Rik, Rdcvf2
          Chromosome Location:
          Chr.13: 51170114 - 51175191 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512580_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nxnl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029173.4 NP_083449.1 1 - 2 596 69
          GenBank mRNA AK015847.1 - 1 - 2 597 69
          BC038905.1 - 1 - 2 599 5025124 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000596.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nxnl2

          Panther Classification:

          Molecular Function -

          oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          sensory perception
          visual perception
          sensory perception of smell
          photoreceptor cell maintenance
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29031119 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29031121 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29031120 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605556 BC038905.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605556 NM_029173.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605556 AK015847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605556 BC038905.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605556 NM_029173.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605556 AK015847.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 7914 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116978 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605556 BC038905.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605556 NM_029173.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605556 AK015847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605556 BC038905.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605556 NM_029173.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605556 AK015847.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29031132 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116978 - -
          Whole Mouse Genome G4122A A_52_P605556 BC038905.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605556 NM_029173.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605556 AK015847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605556 BC038905.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605556 NM_029173.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605556 AK015847.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112236 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116978 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29031121 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29031120 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29031119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29031059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29031123 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10405178 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405176 - -
          Affymetrix HT MG-430 PM Array Plate 1451861_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429133_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10405174 - -
          Mouse Genome 430 2.0 Array 1451861_at - -
          Mouse Genome 430 2.0 Array 1429133_at - -
          Mouse Genome 430 2.0 Array 1451861_PM_at - -
          Mouse Genome 430 2.0 Array 1429133_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75124_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75124_at - -
          Affymetrix Mouse Clariom S Array PSR1300003762.mm.1 NM_029173.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300003762.mm.1 NM_029173.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300003762.mm.1 TC1300000596.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300003763.mm.1 NM_029173.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300003763.mm.1 NM_029173.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300003763.mm.1 TC1300000596.mm.1 MTA TC
          Mouse Expression Set 430 1429133_PM_at - -
          Mouse Expression Set 430 1429133_at - -
          Affymetrix HT Mouse Genome 430A Array 1451861_at - -
          Affymetrix HT Mouse Genome 430A Array 1451861_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429133_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429133_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 746950 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1090520 - -
          Illumina mouse-6 v1.0 expression beadchip 1090520 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090520 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2702547 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2702547 - -
          mouse-6 v1.1 (Illumina) scl44779.2.1_92-S - -
          Illumina Sentrix mouse-6.v1 bead array 1090520 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44779.2.1_92-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090520 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2702547 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2702547 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline