Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512620_m1
          See other CFAP126 GE Assays ›
          Gene Symbol:
          Cfap126
          Gene Name:
          cilia and flagella associated protein 126
          Gene Aliases:
          1700009P17Rik, AV282292, Flattop, Fltp
          Chromosome Location:
          Chr.1: 171113744 - 171126967 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512620_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cfap126
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081275.1 NP_001074744.1 4 - 5 451 75
          GenBank mRNA AK005804.1 - 4 - 5 453 75
          BC061017.1 - 3 - 4 177 6774925 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001577.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell projection organization
          cilium organization
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826098 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826100 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826099 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430259 AK005804.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430259 BC061017.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430259 NM_001081275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430259 AK005804.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430259 BC061017.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430259 NM_001081275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186668 AK005804.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186668 BC061017.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186668 NM_001081275.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45119 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150158 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430259 AK005804.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430259 BC061017.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430259 NM_001081275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430259 AK005804.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430259 BC061017.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430259 NM_001081275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186668 AK005804.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186668 BC061017.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186668 NM_001081275.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150158 - -
          Whole Mouse Genome G4122A A_51_P430259 AK005804.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430259 BC061017.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430259 NM_001081275.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430259 AK005804.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430259 BC061017.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430259 NM_001081275.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150158 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826099 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826100 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20826098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20826147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00062439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01081251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00286343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00268040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00942891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00519957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01086136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00743835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00942894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00306852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00962058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20826217 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351524 - -
          Affymetrix HT MG-430 PM Array Plate 1429181_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10351517 - -
          Mouse Genome 430 2.0 Array 1429181_PM_at - -
          Mouse Genome 430 2.0 Array 1429181_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75472_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75472_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100012897.mm.1 NM_001081275.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012897.mm.1 NM_001081275.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012897.mm.1 TC0100001577.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012896.mm.1 NM_001081275.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012896.mm.1 NM_001081275.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012896.mm.1 TC0100001577.mm.1 MTA TC
          Mouse Expression Set 430 1429181_PM_at - -
          Mouse Expression Set 430 1429181_at - -
          Affymetrix HT Mouse Genome 430B Array 1429181_at - -
          Affymetrix HT Mouse Genome 430B Array 1429181_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2645509 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2645509 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2645509 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline