Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512651_m1
          See other TM4SF5 GE Assays ›
          Gene Symbol:
          Tm4sf5
          Gene Name:
          transmembrane 4 superfamily member 5
          Gene Aliases:
          2010003F10Rik, AI323987
          Chromosome Location:
          Chr.11: 70505274 - 70511183 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512651_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tm4sf5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029360.3 NP_083636.2 1 - 2 232 62
          GenBank mRNA AK008064.1 - 1 - 2 230 62
          BB869909.1 - 1 - 2 233 62
          BC010782.1 - 1 - 2 192 4219239 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000953.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tm4sf5

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163138 BC010782.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163138 NM_029360.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163138 AK008064.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163138 BB798656.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 287001 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2991 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119054 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163138 BC010782.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163138 NM_029360.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163138 AK008064.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163138 BB798656.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27901025 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119054 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07388708 - -
          Whole Mouse Genome G4122A A_51_P119387 BC010782.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119387 NM_029360.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119387 AK008064.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119387 BB798656.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104403 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103609 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09256245 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09729417 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09256236 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09256242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27901139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09662112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09303636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09413529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09299011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09413506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09084595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09349906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09303637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09413525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09413515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09662111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09569181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09413520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09080156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09080154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09734376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09630145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27901035 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10377844 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377845 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377843 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377842 - -
          Affymetrix HT MG-430 PM Array Plate 1424445_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10377841 - -
          Mouse Genome 430 2.0 Array 1424445_at - -
          Mouse Genome 430 2.0 Array 1424445_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75604_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75604_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100008400.mm.1 NM_029360.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008400.mm.1 NM_029360.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008400.mm.1 TC1100000953.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008399.mm.1 NM_029360.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008399.mm.1 NM_029360.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008399.mm.1 TC1100000953.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008401.mm.1 NM_029360.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008401.mm.1 NM_029360.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008401.mm.1 TC1100000953.mm.1 MTA TC
          Mouse Expression Set 430 1424445_at - -
          Mouse Expression Set 430 1424445_PM_at - -
          Murine Genome U74v2 Set 104707_PM_at - -
          Murine Genome U74v2 Set 104707_at - -
          Mouse Genome 430A 2.0 Array 1424445_PM_at - -
          Mouse Genome 430A 2.0 Array 1424445_at - -
          Affymetrix HT Mouse Genome 430A Array 1424445_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424445_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 542331 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3120280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120280 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213662 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213662 - -
          mouse-6 v1.1 (Illumina) scl41337.5.1_185-S - -
          Illumina Sentrix mouse-6.v1 bead array 3120280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41337.5.1_185-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2945330 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2945330 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline