Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512889_m1
          See other TRMT5 GE Assays ›
          Gene Symbol:
          Trmt5
          Gene Name:
          TRM5 tRNA methyltransferase 5
          Gene Aliases:
          2610027O18Rik, mKIAA1393
          Chromosome Location:
          Chr.12: 73268515 - 73286787 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512889_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trmt5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029580.3 NP_083856.1 2 - 3 665 71
          GenBank mRNA AK011576.1 - 2 - 3 666 71
          AK049127.1 - 2 - 3 664 71
          AK220389.1 - 1 - 2 412 71
          BC012521.1 - 2 - 3 666 2648293 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001916.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trmt5

          Panther Classification:

          Molecular Function -

          RNA methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA processing
          biological_process
          tRNA methylation
          methylation
          molecular_function
          methyltransferase activity
          tRNA (guanine-N1-)-methyltransferase activity
          transferase activity
          tRNA (guanine(37)-N(1))-methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536528 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536531 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536527 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536526 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536529 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083967 BC012521.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083967 AK011576.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083967 AK049127.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083967 NM_029580.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960023 BC012521.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913432 AF465352.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913432 AK039558.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913432 AK170258.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913432 BC052502.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913432 BC052734.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913432 BC063052.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913432 NM_153143.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129369 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154134 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083967 BC012521.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083967 AK011576.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083967 AK049127.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083967 NM_029580.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28536492 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154134 - -
          Whole Mouse Genome G4122A A_52_P779660 AK075622.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P421882 BC012521.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P421882 AK011576.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P421882 AK049127.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P421882 NM_029580.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140747 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154134 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10348126 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10348134 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10348135 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10348127 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10348125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536526 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536531 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536528 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536529 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28536527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28536566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10348135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10348137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10021659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10114739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10335180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10114748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10335185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09962776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09863489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28536493 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10400978 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400977 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400976 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400980 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400979 - -
          Affymetrix HT MG-430 PM Array Plate 1452086_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10400975 - -
          Mouse Genome 430 2.0 Array 1452086_PM_at - -
          Mouse Genome 430 2.0 Array 1452086_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76357_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76357_at - -
          Affymetrix Mouse Clariom S Array PSR1200013191.mm.1 NM_029580.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013191.mm.1 NM_029580.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013191.mm.1 TC1200001916.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013192.mm.1 NM_029580.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013192.mm.1 NM_029580.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013192.mm.1 TC1200001916.mm.1 MTA TC
          Mouse Expression Set 430 1452086_PM_at - -
          Mouse Expression Set 430 1452086_at - -
          Mouse Genome 430A 2.0 Array 1452086_at - -
          Mouse Genome 430A 2.0 Array 1452086_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452086_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452086_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 848564 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3390722 - -
          Illumina mouse-6 v1.0 expression beadchip 3390722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003390722 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2616161 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2616161 - -
          mouse-6 v1.1 (Illumina) scl42348.7.1_110-S - -
          Illumina Sentrix mouse-6.v1 bead array 3390722 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42348.7.1_110-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3390722 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2616161 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2616161 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline