Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512911_s1
          See other PBP2 GE Assays ›
          Gene Symbol:
          Pbp2
          Gene Name:
          phosphatidylethanolamine binding protein 2
          Gene Aliases:
          1700023A18Rik, AI326308, Pebp-2, Pebp2
          Chromosome Location:
          Chr.6: 135309129 - 135310384 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512911_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029595.3 NP_083871.3 1 926 63
          GenBank mRNA AF307146.1 - 1 906 63
          AF307147.1 - 2 864 63
          AK006258.1 - 1 953 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003317.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pbp2

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          negative regulation of peptidase activity
          nucleotide binding
          molecular_function
          serine-type endopeptidase inhibitor activity
          ATP binding
          lipid binding
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952482 NM_029595.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70000 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174502 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952482 NM_029595.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24866234 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174502 - -
          Whole Mouse Genome G4122A A_51_P324450 AF307147.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P324450 AK006258.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P324450 NM_029595.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P324450 AF307146.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134939 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174502 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05648863 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05648857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24866245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05121295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05680501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05648860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05392518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05688404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05688406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24866203 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548782 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548783 - -
          Affymetrix HT MG-430 PM Array Plate 1424793_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10548779 - -
          Mouse Genome 430 2.0 Array 1424793_a_at - -
          Mouse Genome 430 2.0 Array 1424793_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76400_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76400_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600026009.mm.1 TC0600003317.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600026009.mm.1 NM_029595.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026009.mm.1 NM_029595.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026011.mm.1 TC0600003317.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600026011.mm.1 NM_029595.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026011.mm.1 NM_029595.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026010.mm.1 TC0600003317.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600026010.mm.1 NM_029595.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026010.mm.1 NM_029595.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026008.mm.1 TC0600003317.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600026008.mm.1 NM_029595.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026008.mm.1 NM_029595.3 Refseq
          Mouse Expression Set 430 1424793_PM_a_at - -
          Mouse Expression Set 430 1424793_a_at - -
          Mouse Genome 430A 2.0 Array 1424793_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1424793_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424793_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424793_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 923391 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 430156 - -
          Illumina mouse-6 v1.0 expression beadchip 430156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000430156 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1250250 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250250 - -
          mouse-6 v1.1 (Illumina) scl28289.1.79_12-S - -
          Illumina Sentrix mouse-6.v1 bead array 430156 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28289.1.79_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 430156 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1250250 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1250250 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0