Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512957_m1
          See other ABHD14B GE Assays ›
          Gene Symbol:
          Abhd14b
          Gene Name:
          abhydrolase domain containing 14b
          Gene Aliases:
          1810013B01Rik
          Chromosome Location:
          Chr.9: 106447737 - 106452923 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512957_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abhd14b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029631.3 NP_083907.3 2 - 3 293 99
          GenBank mRNA AK007469.1 - 2 - 3 248 99
          BC019410.1 - 2 - 3 272 4191819 99
          CJ186593.1 - 2 - 3 293 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001348.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abhd14b

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          positive regulation of transcription from RNA polymerase II promoter
          GPI-anchor transamidase activity
          hydrolase activity
          coenzyme F390-A hydrolase activity
          coenzyme F390-G hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059640 BF461776.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059640 NM_029631.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059640 AK007469.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513928 AK050146.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513928 AK160304.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513928 AK162829.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513928 AK162907.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513928 BC039806.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513928 NM_172383.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110040 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138242 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059640 BF461776.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059640 NM_029631.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059640 AK007469.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803458 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138242 - -
          Whole Mouse Genome G4122A A_51_P270083 BC019410.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270083 BF461776.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270083 NM_029631.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270083 AK007469.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133556 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138242 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08153041 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07961429 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08153040 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08153039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07711263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07554673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08042102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07711251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07961429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08153037 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10588508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588507 - -
          Affymetrix HT MG-430 PM Array Plate 1451326_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10588505 - -
          Mouse Genome 430 2.0 Array 1451326_PM_at - -
          Mouse Genome 430 2.0 Array 1451326_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76491_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76491_at - -
          Affymetrix Mouse Clariom S Array PSR0900010516.mm.1 NM_029631.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010516.mm.1 NM_029631.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010516.mm.1 TC0900001348.mm.1 MTA TC
          Mouse Expression Set 430 1451326_PM_at - -
          Mouse Expression Set 430 1451326_at - -
          Mouse Genome 430A 2.0 Array 1451326_at - -
          Mouse Genome 430A 2.0 Array 1451326_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451326_at - -
          Affymetrix HT Mouse Genome 430A Array 1451326_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 759163 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3800068 - -
          Illumina mouse-6 v1.0 expression beadchip 3800068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003800068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2774244 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2774244 - -
          mouse-6 v1.1 (Illumina) scl36510.4_431-S - -
          Illumina Sentrix mouse-6.v1 bead array 3800068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36510.4_431-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3800068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3007862 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3007862 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774244 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36510.4_431-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline