Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512990_m1
          See other 1700063K16RIK GE Assays ›
          Gene Symbol:
          1700063K16Rik
          Gene Name:
          RIKEN cDNA 1700063K16 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512990_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700063K16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK006873.1 - 5 - 6 514 80

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161503 AK006873.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P161503 AK006873.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58687 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 90581 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161503 AK006873.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P161503 AK006873.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08651202 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08651157 - -
          Whole Mouse Genome G4122A A_51_P161503 AK006873.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P161503 AK006873.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131654 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08651202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12673107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12875047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12682713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12590441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12726676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12596378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12630767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12758743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12880123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30886864 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432422_PM_at - -
          Mouse Genome 430 2.0 Array 1432422_at - -
          Mouse Genome 430 2.0 Array 1432422_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76610_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76610_at - -
          Mouse Expression Set 430 1432422_at - -
          Mouse Expression Set 430 1432422_PM_at - -
          Murine Genome U74v2 Set 170167_PM_at - -
          Murine Genome U74v2 Set 170167_at - -
          Affymetrix HT Mouse Genome 430B Array 1432422_at - -
          Affymetrix HT Mouse Genome 430B Array 1432422_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 729807 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103710068 - -
          Illumina mouse-6 v1.0 expression beadchip 103710068 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2550130 - -
          Illumina Sentrix mouse-6.v1 bead array 103710068 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103710068 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2550130 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline