Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00513246_m1
          See other 5033406O09RIK GE Assays ›
          Gene Symbol:
          5033406O09Rik
          Gene Name:
          RIKEN cDNA 5033406O09 gene
          Gene Aliases:
          9130204G15Rik
          Chromosome Location:
          Chr.12: 111941991 - 111944482 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00513246_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5033406O09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029464.1 2 - 3 506 135
          GenBank mRNA AK017159.1 - 2 - 3 466 135
          AK020272.1 - 2 - 3 233 135
          AK079861.1 - 2 - 3 500 135
          AK134157.1 - 2 - 3 440 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002419.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2324976 AK134157.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2324976 NR_029464.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192334 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2324976 AK134157.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2324976 NR_029464.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365948 AK017159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365948 AK020272.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365948 AK079861.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365948 AK134157.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365948 NR_029464.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10088600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09755771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09960486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09770126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10077160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28747025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09960511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10077161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10398647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28746982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09960501 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430079_PM_at - -
          Mouse Genome 430 2.0 Array 1430079_PM_at - -
          Mouse Genome 430 2.0 Array 1430079_at - -
          Affymetrix Mouse Clariom S Array PSR1200016809.mm.1 NR_029464.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016809.mm.1 NR_029464.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016809.mm.1 TC1200002419.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016811.mm.1 NR_029464.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016811.mm.1 NR_029464.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016811.mm.1 TC1200002419.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016810.mm.1 NR_029464.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016810.mm.1 NR_029464.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016810.mm.1 TC1200002419.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016812.mm.1 NR_029464.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016812.mm.1 NR_029464.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016812.mm.1 TC1200002419.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016814.mm.1 NR_029464.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016814.mm.1 NR_029464.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016814.mm.1 TC1200002419.mm.1 MTA TC
          Mouse Expression Set 430 1430079_PM_at - -
          Mouse Expression Set 430 1430079_at - -
          Affymetrix HT Mouse Genome 430B Array 1430079_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430079_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 753864 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106450048 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline