Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00513354_m1
          See other HHIPL2 GE Assays ›
          Gene Symbol:
          Hhipl2
          Gene Name:
          hedgehog interacting protein-like 2
          Gene Aliases:
          4930507C10Rik, AW208802, Hhip3
          Chromosome Location:
          Chr.1: 183413929 - 183437053 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00513354_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hhipl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK019671.1 - 6 - 7 1545 66
          BC034362.1 - 5 - 6 1290 3154701 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001757.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 BC034362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 NM_030175.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 AK019671.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 BC034362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 NM_030175.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344364 AK019671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 BC034362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 NM_030175.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 AK019671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 BC034362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 NM_030175.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344364 AK019671.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 BC034362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 NM_030175.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 AK019671.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 BC034362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 NM_030175.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344364 AK019671.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116097 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10598135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598134 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598133 - -
          Affymetrix HT MG-430 PM Array Plate 1429157_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10598126 - -
          Mouse Genome 430 2.0 Array 1429157_PM_at - -
          Mouse Genome 430 2.0 Array 1429157_at - -
          Affymetrix Mouse Clariom S Array PSR0100014702.mm.1 NM_030175.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0100014702.mm.1 TC0100001757.mm.1 MTA TC
          Mouse Expression Set 430 1429157_PM_at - -
          Mouse Expression Set 430 1429157_at - -
          Affymetrix HT Mouse Genome 430B Array 1429157_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429157_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 445119 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7000026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620374 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620374 - -
          mouse-6 v1.1 (Illumina) scl078772.9_102-S - -
          Illumina Sentrix mouse-6.v1 bead array 7000026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl078772.9_102-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000026 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2620374 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620374 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline