Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00513626_g1
          See other 6330403K07RIK GE Assays ›
          Gene Symbol:
          6330403K07Rik
          Gene Name:
          RIKEN cDNA 6330403K07 gene
          Gene Aliases:
          AI415455, AV161034, UGS148
          Chromosome Location:
          Chr.11: 71031941 - 71033617 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00513626_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6330403K07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134022.2 NP_598783.1 1 - 2 67
          GenBank mRNA AJ271140.1 - 1 - 2 67
          AK018106.1 - 1 - 2 67
          AK083923.1 - 1 - 2 67
          BC069976.1 - 1 - 2 970 6518302 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003122.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 6330403K07Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27904028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27904030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27904029 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27904027 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417251 AK018106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417251 AK083923.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417251 BC069976.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417251 NM_134022.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417251 AJ271140.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417251 AK018106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417251 AK083923.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417251 BC069976.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417251 NM_134022.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417251 AJ271140.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553365 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121939 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417251 AK018106.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417251 AK083923.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417251 BC069976.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417251 NM_134022.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417251 AJ271140.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417251 AK018106.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417251 AK083923.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417251 BC069976.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417251 NM_134022.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417251 AJ271140.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904032 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121939 - -
          Whole Mouse Genome G4122A A_51_P417251 AK018106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417251 AK083923.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417251 BC069976.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417251 NM_134022.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417251 AJ271140.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417251 AK018106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417251 AK083923.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417251 BC069976.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417251 NM_134022.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417251 AJ271140.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136362 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121939 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09469502 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09469500 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09004791 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09004798 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27904029 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27904030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27904027 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27904028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09376254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09275252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09004799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09099807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09004798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09412619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09225615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09205493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904033 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388044 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388043 - -
          Affymetrix HT MG-430 PM Array Plate 1426766_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10388042 - -
          Mouse Genome 430 2.0 Array 1426766_at - -
          Mouse Genome 430 2.0 Array 1426766_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 103712_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 103712_at - -
          Affymetrix Mouse Clariom S Array PSR1100028933.mm.1 NM_134022.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028933.mm.1 NM_134022.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028933.mm.1 TC1100003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028932.mm.1 NM_134022.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028932.mm.1 NM_134022.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028932.mm.1 TC1100003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028935.mm.1 NM_134022.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028935.mm.1 NM_134022.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028935.mm.1 TC1100003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028934.mm.1 NM_134022.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028934.mm.1 NM_134022.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028934.mm.1 TC1100003122.mm.1 MTA TC
          Mouse Expression Set 430 1426766_at - -
          Mouse Expression Set 430 1426766_PM_at - -
          Murine Genome U74v2 Set 161993_r_at - -
          Murine Genome U74v2 Set 95559_PM_at - -
          Murine Genome U74v2 Set 161993_PM_r_at - -
          Murine Genome U74v2 Set 95559_at - -
          Murine Genome U74v2 Set 161354_PM_f_at - -
          Murine Genome U74v2 Set 161354_f_at - -
          Murine Genome U74v2 Set 169989_PM_r_at - -
          Murine Genome U74v2 Set 169989_r_at - -
          Mouse Genome 430A 2.0 Array 1426766_at - -
          Mouse Genome 430A 2.0 Array 1426766_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426766_at - -
          Affymetrix HT Mouse Genome 430A Array 1426766_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 446228 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2060504 - -
          Illumina mouse-6 v1.0 expression beadchip 2060504 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002060504 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2594139 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2594139 - -
          mouse-6 v1.1 (Illumina) scl0103712.1_49-S - -
          Illumina Sentrix mouse-6.v1 bead array 2060504 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0103712.1_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060504 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2594139 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594139 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0103712.1_49-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline