Search
Search

Cabp4| Gene Symbol: Cabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144532.2 | NP_653115.1 | 4 - 5 | 690 | 59 | |
| GenBank mRNA | AK010631.1 | - | 1 - 2 | 232 | 59 | |
| AY039218.1 | - | 4 - 5 | 685 | 59 | ||
| BC029002.1 | - | 3 - 4 | 1930 | 59 | ||
| BC049263.1 | - | 4 - 5 | 690 | 5366999 | 59 | |
| BC053330.1 | - | 3 - 4 | 1977 | 59 | ||
Set Membership: |
|