Search
Search

Slamf7| Gene Symbol: Slamf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144539.5 | NP_653122.2 | 3 - 4 | 815 | 76 | |
| GenBank mRNA | AB196816.1 | - | 2 - 3 | 362 | 76 | |
| AB196817.1 | - | 2 - 3 | 362 | 76 | ||
| AB196818.1 | - | 2 - 3 | 362 | 76 | ||
| AB196819.1 | - | 2 - 3 | 362 | 76 | ||
| AB196837.1 | - | 2 - 3 | 362 | 76 | ||
| AF467909.1 | - | 2 - 3 | 76 | |||
| AF467910.1 | - | 2 - 3 | 362 | 76 | ||
| AF467911.1 | - | 2 - 3 | 362 | 76 | ||
| AK016183.1 | - | 3 - 4 | 619 | 76 | ||
| AK030135.1 | - | 3 - 4 | 676 | 76 | ||
| AK030148.1 | - | 3 - 4 | 816 | 76 | ||
| AK040678.1 | - | 2 - 3 | 443 | 76 | ||
| AK089525.1 | - | 2 - 3 | 443 | 76 | ||
| AK172665.1 | - | 2 - 3 | 409 | 76 | ||
| BC011154.1 | - | 2 - 3 | 374 | 4168309 | 76 | |
Set Membership: |
|