Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00513848_m1
          See other NDUFAF7 GE Assays ›
          Gene Symbol:
          Ndufaf7
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 7
          Gene Aliases:
          2410091C18Rik, AL033374, PRO1853
          Chromosome Location:
          Chr.17: 78937106 - 78948052 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00513848_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufaf7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028611.3 NP_082887.2 8 - 9 954 77
          GenBank mRNA AK010752.1 - 8 - 9 953 77
          AK090097.1 - 8 - 9 953 77
          BC028524.1 - 1 - 2 193 77
          BC108350.1 - 8 - 9 941 4220568 77
          BC116213.1 - 1 - 2 46 77
          BC116214.1 - 1 - 2 46 77
          BC128473.1 - 1 - 2 46 77
          BC128474.1 - 1 - 2 46 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001192.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC028524.1,BC116213.1,BC116214.1,BC128473.1,BC128474.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufaf7

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial respiratory chain complex I assembly
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180826 BC108350.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180826 BC116213.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180826 BC116214.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180826 BC128473.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180826 BC128474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180826 BC108350.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180826 BC116213.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180826 BC116214.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180826 BC128473.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P180826 BC128474.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463889 AK038863.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463889 AK052153.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463889 AK090131.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463889 AK122562.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463889 AK147808.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463889 AK147897.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463889 AK170702.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463889 BC039227.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 80655 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131422 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145260 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180826 BC108350.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180826 BC116213.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180826 BC116214.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180826 BC128473.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180826 BC128474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180826 BC108350.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180826 BC116213.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180826 BC116214.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180826 BC128473.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P180826 BC128474.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145260 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131422 - -
          Whole Mouse Genome G4122A A_52_P406459 BC108350.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P406459 CX236822.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P406459 AK090097.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P406459 NM_028611.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180826 BC108350.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180826 BC116213.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180826 BC116214.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180826 BC128473.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180826 BC128474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180826 BC108350.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180826 BC116213.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180826 BC116214.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180826 BC128473.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P180826 BC128474.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116268 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125412 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131422 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31374593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13122876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13583255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13294903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13283073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13122878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13548517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13348592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13453203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13294901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374550 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10447052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447049 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447051 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447046 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447048 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447045 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447047 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447044 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447043 - -
          Affymetrix HT MG-430 PM Array Plate 1427904_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10447042 - -
          Mouse Genome 430 2.0 Array 1427904_PM_s_at - -
          Mouse Genome 430 2.0 Array 1427904_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73694_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73694_at - -
          Affymetrix Mouse Clariom S Array PSR1700011275.mm.1 NM_028611.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011275.mm.1 NM_028611.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011275.mm.1 TC1700001192.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700011272.mm.1 NM_028611.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011272.mm.1 NM_028611.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011272.mm.1 TC1700001192.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700011274.mm.1 NM_028611.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011274.mm.1 NM_028611.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011274.mm.1 TC1700001192.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700011273.mm.1 TC1700001192.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700011276.mm.1 NM_028611.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011276.mm.1 NM_028611.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700011276.mm.1 TC1700001192.mm.1 MTA TC
          Mouse Expression Set 430 1427904_s_at - -
          Mouse Expression Set 430 1427904_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1427904_s_at - -
          Mouse Genome 430A 2.0 Array 1427904_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427904_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427904_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 492890 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5720017 - -
          MouseWG-6 v2 Expression BeadChip 360369 - -
          Illumina mouse-6 v1.0 expression beadchip 360369 - -
          Illumina mouse-6 v1.0 expression beadchip 5720017 - -
          Illumina mouse-6 v1.0 expression beadchip 6110114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000360369 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006110114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005720017 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2618048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2598520 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2675254 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2618048 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2675254 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2598520 - -
          mouse-6 v1.1 (Illumina) scl0001759.1_49-S - -
          mouse-6 v1.1 (Illumina) scl0001748.1_173-S - -
          mouse-6 v1.1 (Illumina) scl073694.3_57-S - -
          Illumina Sentrix mouse-6.v1 bead array 360369 - -
          Illumina Sentrix mouse-6.v1 bead array 6110114 - -
          Illumina Sentrix mouse-6.v1 bead array 5720017 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001748.1_173-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl073694.3_57-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001759.1_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360369 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110114 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720017 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2598520 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2618048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2675254 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2618048 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2675254 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2598520 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001759.1_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl073694.3_57-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline