Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00514030_m1
          See other CD3EAP GE Assays ›
          Gene Symbol:
          Cd3eap
          Gene Name:
          CD3E antigen, epsilon polypeptide associated protein
          Gene Aliases:
          1190028F09, 2610103M17Rik, ASE-1, Ase1, CAST, PAF49
          Chromosome Location:
          Chr.7: 19356007 - 19359483 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00514030_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd3eap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145822.2 NP_665821.1 2 - 3 239 67
          GenBank mRNA AB091121.1 - 2 - 3 176 67
          AK011811.1 - 2 - 3 212 67
          BC016115.1 - 2 - 3 204 67
          BC029206.1 - 2 - 3 187 4039808 67
          BC071199.1 - 2 - 3 190 6478245 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002479.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd3eap

          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA transcription
          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24992865 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24992866 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24992859 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24992858 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24992860 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317443 BC071199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317443 BC016115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317443 BC029206.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317443 NM_145822.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317443 AB091121.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317443 BC071199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317443 BC016115.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317443 BC029206.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317443 NM_145822.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317443 AB091121.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681039 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137897 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317443 BC071199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317443 BC016115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317443 BC029206.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317443 NM_145822.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317443 AB091121.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317443 BC071199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317443 BC016115.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317443 BC029206.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317443 NM_145822.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317443 AB091121.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317443 BC071199.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317443 BC016115.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317443 BC029206.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317443 NM_145822.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317443 AB091121.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317443 BC071199.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317443 BC016115.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317443 BC029206.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317443 NM_145822.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P317443 AB091121.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24992861 - -
          Whole Mouse Genome G4122A A_51_P317443 BC071199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317443 BC016115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317443 BC029206.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317443 NM_145822.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317443 AB091121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317443 BC071199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317443 BC016115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317443 BC029206.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317443 NM_145822.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317443 AB091121.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118265 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06592637 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06592635 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06592633 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06070558 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06592638 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06592644 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06592630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06070562 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06592634 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24992858 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24992866 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24992860 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24992859 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24992865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24992868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06562409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06592648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24992848 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560489 - -
          Affymetrix HT MG-430 PM Array Plate 1430138_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550650 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560487 - -
          Mouse Genome 430 2.0 Array 1430138_PM_at - -
          Mouse Genome 430 2.0 Array 1430138_at - -
          Affymetrix Mouse Clariom S Array PSR0700021807.mm.1 TC0700002479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021807.mm.1 NM_145822.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021807.mm.1 NM_145822.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021805.mm.1 TC0700002479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021805.mm.1 NM_145822.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021805.mm.1 NM_145822.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021810.mm.1 TC0700002479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021810.mm.1 NM_145822.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021810.mm.1 NM_145822.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021809.mm.1 TC0700002479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021809.mm.1 NM_145822.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021809.mm.1 NM_145822.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021812.mm.1 TC0700002479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021812.mm.1 NM_145822.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021812.mm.1 NM_145822.2 Refseq
          Mouse Expression Set 430 1430138_PM_at - -
          Mouse Expression Set 430 1430138_at - -
          Affymetrix HT Mouse Genome 430B Array 1430138_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430138_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 862993 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 870092 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000870092 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246695 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246695 - -
          mouse-6 v1.1 (Illumina) scl070333.1_111-S - -
          Illumina Sentrix mouse-6.v1 bead array 870092 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl070333.1_111-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 870092 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246695 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246695 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl070333.1_111-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline