Search
Search
| Gene Symbol: Cttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252572.1 | NP_001239501.1 | 5 - 6 | 633 | 64 | |
| NM_007803.5 | NP_031829.2 | 5 - 6 | 633 | 64 | ||
| GenBank mRNA | AK084249.1 | - | 5 - 6 | 443 | 64 | |
| AK146856.1 | - | 5 - 6 | 440 | 64 | ||
| AK148010.1 | - | 5 - 6 | 436 | 64 | ||
| AK165555.1 | - | 5 - 6 | 436 | 64 | ||
| AK168699.1 | - | 5 - 6 | 434 | 64 | ||
| AK168871.1 | - | 5 - 6 | 434 | 64 | ||
| AK168945.1 | - | 5 - 6 | 444 | 64 | ||
| AK197295.1 | - | 1 - 2 | 38 | 64 | ||
| BC011434.1 | - | 5 - 6 | 426 | 3981559 | 64 | |
| BC117820.1 | - | 4 - 5 | 334 | 64 | ||
| U03184.1 | - | 5 - 6 | 422 | 64 | ||
Set Membership: |
|