Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00514704_m1
          See other EMD GE Assays ›
          Gene Symbol:
          Emd
          Gene Name:
          emerin
          Gene Aliases:
          AW550900, Sta
          Chromosome Location:
          Chr.X: 74254673 - 74261563 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00514704_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Emd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007927.3 NP_031953.1 4 - 5 659 88
          GenBank mRNA AK146361.1 - 4 - 5 505 88
          AK160531.1 - 4 - 5 504 87
          AK167852.1 - 4 - 5 502 88
          BC132131.1 - 4 - 5 471 40130408 88
          BC132133.1 - 4 - 5 471 40130410 88
          U73902.1 - 4 - 5 509 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000703.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Emd

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of catenin import into nucleus
          skeletal muscle cell differentiation
          positive regulation of protein export from nucleus
          negative regulation of fibroblast proliferation
          regulation of canonical Wnt signaling pathway
          cellular response to growth factor stimulus
          actin binding
          protein binding
          beta-tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467041 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467102 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467033 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467107 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467040 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467036 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467042 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467035 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467105 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467104 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467101 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467043 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467037 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984337 AK167852.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984337 U73902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984337 NM_007927.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984337 AK146361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984337 AK160531.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984333 AK168209.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984333 BC132131.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984333 BC132133.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984333 U73902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984333 NM_007927.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653406 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100454 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984333 AK168209.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984333 BC132131.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984333 BC132133.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984333 U73902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984333 NM_007927.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984337 AK167852.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984337 U73902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984337 NM_007927.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984337 AK146361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984337 AK160531.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32467078 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100454 - -
          Whole Mouse Genome G4122A A_51_P216147 AK167852.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P216147 U73902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P216147 NM_007927.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P216147 AK146361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P216147 AK160531.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108049 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100454 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15000971 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467101 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467102 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467105 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467039 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467037 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467042 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467041 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467035 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467040 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467043 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467107 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467104 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467033 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32467036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32467067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14925992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15000952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14859080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14565060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15000950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14879338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14529513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32467120 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10600342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600346 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600344 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600345 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600347 - -
          Affymetrix HT MG-430 PM Array Plate 1417357_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10600341 - -
          Mouse Genome 430 2.0 Array 1417357_at - -
          Mouse Genome 430 2.0 Array 1417357_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13726_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13726_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00004662.mm.1 NM_007927.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00004662.mm.1 TC0X00000703.mm.1 MTA TC
          Mouse Expression Set 430 1417357_PM_at - -
          Mouse Expression Set 430 1417357_at - -
          Murine Genome U74v2 Set 103420_at - -
          Murine Genome U74v2 Set 103420_PM_at - -
          Murine Genome U74v2 Set 139263_at - -
          Murine Genome U74v2 Set 139263_PM_at - -
          Mouse Genome 430A 2.0 Array 1417357_at - -
          Mouse Genome 430A 2.0 Array 1417357_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417357_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417357_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 463384 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6520731 - -
          Illumina mouse-6 v1.0 expression beadchip 2230278 - -
          Illumina mouse-6 v1.0 expression beadchip 6520731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2745259 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2630593 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2745259 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2630593 - -
          mouse-6 v1.1 (Illumina) scl54837.5.1_4-S - -
          mouse-6 v1.1 (Illumina) scl0002880.1_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520731 - -
          Illumina Sentrix mouse-6.v1 bead array 2230278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54837.5.1_4-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002880.1_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520731 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2630593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2745259 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2630593 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2745259 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl54837.5.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0002880.1_15-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline