Search
Search

Grid2| Gene Symbol: Grid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008167.2 | NP_032193.1 | 12 - 13 | 1994 | 77 | |
| GenBank mRNA | BC139823.1 | - | 12 - 13 | 1998 | 40141838 | 77 |
| D13266.1 | - | 12 - 13 | 1994 | 77 | ||
Set Membership: |
|