Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00515420_m1
          See other CD19 GE Assays ›
          Gene Symbol:
          Cd19
          Gene Name:
          CD19 antigen
          Gene Aliases:
          AW495831
          Chromosome Location:
          Chr.7: 126408448 - 126415140 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00515420_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009844.2 NP_033974.2 4 - 5 860 69
          GenBank mRNA AK089835.1 - 4 - 5 859 69
          AK163608.1 - 2 - 3 1357 69
          AK172008.1 - 4 - 5 822 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004277.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus Mouse Immune Response 1700 Inventoried Mouse_Immune_Response Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd19

          Gene Ontology Categories:

          Function(s)

          chromosome breakage
          B cell receptor signaling pathway
          positive regulation of release of sequestered calcium ion into cytosol

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079079 AK156125.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079079 AK163608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079079 NM_009844.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079079 AK089835.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378223 AK006482.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378223 BC048662.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763440 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142325 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079079 AK156125.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079079 AK163608.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079079 NM_009844.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079079 AK089835.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P318966 BE686865.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P318966 AK156125.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P318966 AK163608.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P318966 NM_009844.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500376 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142325 - -
          Whole Mouse Genome G4122A A_51_P318966 BE686865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P318966 AK156125.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P318966 AK163608.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P318966 NM_009844.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129670 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121386 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142325 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05901085 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06443343 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06443346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05987937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06152657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05908959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05952123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06152659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05987925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05987935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06443346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06443343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05987936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06150056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06410366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06342013 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10567870 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567875 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567874 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567866 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567872 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567871 - -
          Affymetrix HT MG-430 PM Array Plate 1450570_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10567863 - -
          Mouse Genome 430 2.0 Array 1450570_a_at - -
          Mouse Genome 430 2.0 Array 1450570_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12478_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12478_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700035643.mm.1 NM_009844.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035643.mm.1 NM_009844.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035643.mm.1 TC0700004277.mm.1 MTA TC
          Mouse Expression Set 430 1450570_a_at - -
          Mouse Expression Set 430 1450570_PM_a_at - -
          Murine Genome U74v2 Set 99945_PM_at - -
          Murine Genome U74v2 Set 99945_at - -
          Mouse Genome 430A 2.0 Array 1450570_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1450570_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450570_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450570_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 520081 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6940692 - -
          Illumina mouse-6 v1.0 expression beadchip 6940692 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940692 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228448 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228448 - -
          mouse-6 v1.1 (Illumina) scl30689.14.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940692 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30689.14.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940692 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228448 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2975994 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2975994 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228448 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline