Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00515877_g1
          See other GNG8 GE Assays ›
          Gene Symbol:
          Gng8
          Gene Name:
          guanine nucleotide binding protein (G protein), gamma 8
          Gene Aliases:
          G(y)8
          Chromosome Location:
          Chr.7: 16891737 - 16895451 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00515877_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gng8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010320.3 NP_034450.1 3 - 4 717 88
          GenBank mRNA AK144609.1 - 3 - 4 718 88
          BQ174809.1 - 1 - 2 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000241.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gng8

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          social behavior
          nose development
          cellular response to pheromone
          signal transducer activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983831 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983773 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983830 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983771 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983770 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240136 AK144609.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240136 BC111437.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240136 NM_010320.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240136 BQ174809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240136 AK144609.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240136 BC111437.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240136 NM_010320.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240136 BQ174809.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 820919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254941 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109060 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240136 AK144609.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240136 BC111437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240136 NM_010320.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240136 BQ174809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240136 AK144609.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240136 BC111437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240136 NM_010320.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240136 BQ174809.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240136 AK144609.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240136 BC111437.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240136 NM_010320.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240136 BQ174809.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240136 AK144609.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240136 BC111437.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240136 NM_010320.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240136 BQ174809.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983820 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109060 - -
          Whole Mouse Genome G4122A A_51_P240136 AK144609.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240136 BC111437.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240136 NM_010320.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240136 BQ174809.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240136 AK144609.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240136 BC111437.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240136 NM_010320.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240136 BQ174809.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106437 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109060 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05915245 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05915230 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05999588 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05915242 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05915235 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05915243 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05915237 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983770 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983831 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983773 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983830 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05915232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06414850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05915236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06227767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06610393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06414846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06460285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06460286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06498584 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550390 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550391 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550393 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550392 - -
          Affymetrix HT MG-430 PM Array Plate 1457755_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550388 - -
          Mouse Genome 430 2.0 Array 1457755_PM_at - -
          Mouse Genome 430 2.0 Array 1457755_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14709_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14709_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700001824.mm.1 TC0700000241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001824.mm.1 NM_010320.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001824.mm.1 NM_010320.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001825.mm.1 TC0700000241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001825.mm.1 NM_010320.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001825.mm.1 NM_010320.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001827.mm.1 TC0700000241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001827.mm.1 NM_010320.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001827.mm.1 NM_010320.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001829.mm.1 TC0700000241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001829.mm.1 NM_010320.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001829.mm.1 NM_010320.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001830.mm.1 TC0700000241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001830.mm.1 NM_010320.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001830.mm.1 NM_010320.3 Refseq
          Mouse Expression Set 430 1457755_at - -
          Mouse Expression Set 430 1457755_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457755_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457755_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 797198 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4920358 - -
          Illumina mouse-6 v1.0 expression beadchip 4920358 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004920358 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259653 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259653 - -
          mouse-6 v1.1 (Illumina) scl33043.5.1_34-S - -
          Illumina Sentrix mouse-6.v1 bead array 4920358 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33043.5.1_34-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920358 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1259653 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259653 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33043.5.1_34-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline