Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00515905_s1
          See other HIST1H1T GE Assays ›
          Gene Symbol:
          Hist1h1t
          Gene Name:
          histone cluster 1, H1t
          Gene Aliases:
          H1.6, H1ft, H1t
          Chromosome Location:
          Chr.13: 23695811 - 23696545 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00515905_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist1h1t
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010377.3 NP_034507.2 1 468 69
          GenBank mRNA BC119305.1 - 1 8734054 69
          BC119307.1 - 1 8734056 69
          BV210386.1 - 1 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hist1h1t

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          multicellular organism development
          spermatogenesis
          binding of sperm to zona pellucida
          cell differentiation
          sperm motility
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P312615 BC119305.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P312615 BC119307.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P312615 BC119305.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P312615 BC119307.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P312615 BC119305.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P312615 BC119307.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P312615 BC119305.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P312615 BC119307.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P312615 BC119305.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P312615 BC119307.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P312615 BC119305.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P312615 BC119307.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882919 - -
          Whole Mouse Genome G4122A A_51_P312615 BC119305.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312615 BC119307.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312615 BC119305.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312615 BC119307.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129333 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10977895 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10977877 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10977888 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10977876 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10977892 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10977881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10977877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10711625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10977881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10977892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10977875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11100040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10977876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10711624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882908 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404056 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404057 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404058 - -
          Affymetrix HT MG-430 PM Array Plate 1450608_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10404055 - -
          Mouse Genome 430 2.0 Array 1450608_PM_at - -
          Mouse Genome 430 2.0 Array 1450608_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107970_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107970_at - -
          Affymetrix Mouse Clariom S Array PSR1300001434.mm.1 NM_010377.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001434.mm.1 TC1300000256.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001436.mm.1 NM_010377.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001436.mm.1 TC1300000256.mm.1 MTA TC
          Mouse Expression Set 430 1450608_PM_at - -
          Mouse Expression Set 430 1450608_at - -
          Murine Genome U74v2 Set 100280_PM_at - -
          Murine Genome U74v2 Set 100280_at - -
          Mouse Genome 430A 2.0 Array 1450608_at - -
          Mouse Genome 430A 2.0 Array 1450608_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450608_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450608_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 377945 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 130632 - -
          Illumina mouse-6 v1.0 expression beadchip 130632 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000130632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2644569 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2644569 - -
          mouse-6 v1.1 (Illumina) scl12331.1.1_225-S - -
          Illumina Sentrix mouse-6.v1 bead array 130632 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl12331.1.1_225-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 130632 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2644569 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2644569 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0