Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00516104_m1
          See other KLF4 GE Assays ›
          Gene Symbol:
          Klf4
          Gene Name:
          Kruppel-like factor 4 (gut)
          Gene Aliases:
          EZF, Gklf, Zie
          Chromosome Location:
          Chr.4: 55527137 - 55532475 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00516104_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010637.3 NP_034767.2 2 - 3 735 77
          GenBank mRNA AK144942.1 - 2 - 3 733 77
          AK155343.1 - 2 - 3 733 77
          BC010301.1 - 2 - 3 597 3156339 77
          CF163360.1 - 1 - 2 79 77
          JF277566.1 - 1 - 2 131 77
          U20344.1 - 2 - 3 441 77
          U70662.1 - 2 - 3 729 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002694.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klf4

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          negative regulation of cell proliferation
          epidermal cell differentiation
          response to organic substance
          negative regulation of phosphatidylinositol 3-kinase signaling
          negative regulation of muscle hyperplasia
          stem cell population maintenance
          cell differentiation
          post-embryonic camera-type eye development
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of cellular protein metabolic process
          response to retinoic acid
          negative regulation of heterotypic cell-cell adhesion
          post-embryonic hemopoiesis
          regulation of cell proliferation
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of phosphatidylinositol 3-kinase activity
          negative regulation of interleukin-8 biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          fat cell differentiation
          regulation of cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of hemoglobin biosynthetic process
          negative regulation of smooth muscle cell proliferation
          epidermis morphogenesis
          positive regulation of protein metabolic process
          negative regulation of protein kinase B signaling
          positive regulation of telomerase activity
          canonical Wnt signaling pathway
          negative regulation of response to cytokine stimulus
          negative regulation of ERK1 and ERK2 cascade
          cellular response to growth factor stimulus
          cellular response to laminar fluid shear stress
          negative regulation of cell migration involved in sprouting angiogenesis
          negative regulation of chemokine (C-X-C motif) ligand 2 production
          core promoter proximal region sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding transcription factor recruiting
          core promoter binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          transcription cofactor binding
          nucleic acid binding
          DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          beta-catenin binding
          transcription factor binding
          phosphatidylinositol 3-kinase regulator activity
          histone deacetylase binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400021741.mm.1 TC0400002694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021741.mm.1 NM_010637.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021741.mm.1 NM_010637.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021740.mm.1 TC0400002694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021740.mm.1 NM_010637.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021740.mm.1 NM_010637.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021742.mm.1 TC0400002694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021742.mm.1 NM_010637.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021742.mm.1 NM_010637.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 440920 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline