Search
Search

Cntn2| Gene Symbol: Cntn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177129.5 | NP_796103.2 | 16 - 17 | 2429 | 57 | |
| GenBank mRNA | AK147573.1 | - | 15 - 16 | 2557 | 57 | |
| AK147602.1 | - | 16 - 17 | 2430 | 57 | ||
| AK147658.1 | - | 15 - 16 | 2557 | 57 | ||
| AK147673.1 | - | 15 - 16 | 2582 | 57 | ||
| BC053033.1 | - | 5 - 6 | 637 | 57 | ||
| BC066106.1 | - | 16 - 17 | 2349 | 30362931 | 57 | |
| BC079585.1 | - | 2 - 3 | 205 | 57 | ||
| X81365.1 | - | 1 - 2 | 137 | 57 | ||
Set Membership: |
|