Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00516381_m1
          See other GNB3 GE Assays ›
          Gene Symbol:
          Gnb3
          Gene Name:
          guanine nucleotide binding protein (G protein), beta 3
          Gene Aliases:
          Gnb-3
          Chromosome Location:
          Chr.6: 124834240 - 124840275 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00516381_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013530.1 NP_038558.1 3 - 4 483 79
          GenBank mRNA AK014789.1 - 3 - 4 202 79
          AK030354.1 - 2 - 3 424 79
          BC018239.1 - 3 - 4 483 4511683 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003121.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK030354.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gnb3

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          GTPase activity
          signal transducer activity
          spectrin binding
          GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293862 BC018239.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293862 NM_013530.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293862 AK014789.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293862 AK030354.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293862 BC018239.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293862 NM_013530.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293862 AK014789.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P293862 AK030354.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75161 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61768 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100959 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293862 BC018239.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293862 NM_013530.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293862 AK014789.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293862 AK030354.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293862 BC018239.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293862 NM_013530.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293862 AK014789.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P293862 AK030354.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293862 BC018239.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293862 NM_013530.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293862 AK014789.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293862 AK030354.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293862 BC018239.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293862 NM_013530.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293862 AK014789.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P293862 AK030354.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817247 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100959 - -
          Whole Mouse Genome G4122A A_51_P293862 BC018239.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293862 NM_013530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293862 AK014789.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293862 AK030354.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293862 BC018239.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293862 NM_013530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293862 AK014789.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293862 AK030354.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127133 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100959 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05499812 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05825231 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05825241 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05825227 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05825239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05825236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05685583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05685589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05691361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05155632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05155645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05825239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05825227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05685591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05685585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05764165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05825241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05685588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05825231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05155641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24817276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05825235 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10547860 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547859 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547866 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547861 - -
          Affymetrix HT MG-430 PM Array Plate 1449159_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10547858 - -
          Mouse Genome 430 2.0 Array 1449159_PM_at - -
          Mouse Genome 430 2.0 Array 1449159_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14695_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14695_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600024294.mm.1 NM_013530.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024294.mm.1 NM_013530.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024294.mm.1 TC0600003121.mm.1 MTA TC
          Mouse Expression Set 430 1449159_PM_at - -
          Mouse Expression Set 430 1449159_at - -
          Murine Genome U74v2 Set 99238_PM_at - -
          Murine Genome U74v2 Set 99238_at - -
          Mouse Genome 430A 2.0 Array 1449159_at - -
          Mouse Genome 430A 2.0 Array 1449159_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449159_at - -
          Affymetrix HT Mouse Genome 430A Array 1449159_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 758889 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1230167 - -
          Illumina mouse-6 v1.0 expression beadchip 1230167 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001230167 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218235 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218235 - -
          mouse-6 v1.1 (Illumina) scl014695.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 1230167 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014695.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1230167 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218235 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218235 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline