Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00516563_m1
          See other CASP9 GE Assays ›
          Gene Symbol:
          Casp9
          Gene Name:
          caspase 9
          Gene Aliases:
          AI115399, APAF-3, AW493809, CASP-9, Caspase-9, ICE-LAP6, Mch6
          Chromosome Location:
          Chr.4: 141793612 - 141815978 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00516563_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Casp9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001277932.1 NP_001264861.1 1 - 2 377 68
          NM_015733.5 NP_056548.2 1 - 2 377 68
          GenBank mRNA AB019600.1 - 1 - 2 134 68
          AB019601.1 - 1 - 2 134 68
          AK085095.1 - 1 - 2 166 68
          AK085152.1 - 1 - 2 133 68
          AK085561.1 - 1 - 2 152 68
          AK154495.1 - 1 - 2 147 68
          AK156500.1 - 1 - 2 280 68
          AK162863.1 - 1 - 2 157 68
          BC056372.1 - 1 - 2 159 6834067 68
          BC056447.1 - 1 - 2 377 5721188 68
          CT010400.1 - 1 - 2 134 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001801.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Casp9

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular response to DNA damage stimulus
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to UV
          response to organic cyclic compound
          cellular response to UV
          signal transduction in response to DNA damage
          regulation of apoptotic process
          positive regulation of apoptotic process
          positive regulation of neuron apoptotic process
          cellular response to dexamethasone stimulus
          regulation of response to DNA damage stimulus
          cysteine-type endopeptidase activity
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          SH3 domain binding
          protein kinase binding
          cysteine-type endopeptidase activity involved in apoptotic signaling pathway

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400014947.mm.1 NM_001277932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014947.mm.1 NM_015733.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014947.mm.1 TC0400001801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014950.mm.1 NM_001277932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014950.mm.1 NM_015733.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014950.mm.1 TC0400001801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014944.mm.1 NM_001277932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014944.mm.1 NM_015733.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014944.mm.1 TC0400001801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014951.mm.1 NM_001277932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014951.mm.1 NM_015733.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014951.mm.1 TC0400001801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014945.mm.1 NM_001277932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014945.mm.1 NM_015733.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014945.mm.1 TC0400001801.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 401897 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0