Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00516884_m1
          See other HP GE Assays ›
          Gene Symbol:
          Hp
          Gene Name:
          haptoglobin
          Gene Aliases:
          HP-1, preHP2
          Chromosome Location:
          Chr.8: 109575128 - 109579172 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00516884_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017370.2 NP_059066.1 4 - 5 424 69
          GenBank mRNA AK150430.1 - 4 - 5 317 69
          AK150830.1 - 4 - 5 317 69
          AK152295.1 - 4 - 5 317 69
          AK152698.1 - 4 - 5 317 69
          AK159676.1 - 4 - 5 316 69
          BC138872.1 - 4 - 5 299 8861894 69
          M96827.1 - 4 - 5 314 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002905.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hp

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          immune system process
          proteolysis
          acute-phase response
          Notch signaling pathway
          positive regulation of cell death
          response to hydrogen peroxide
          defense response to bacterium
          negative regulation of oxidoreductase activity
          negative regulation of hydrogen peroxide catabolic process
          serine-type endopeptidase activity
          antioxidant activity
          hemoglobin binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048607 M96827.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048607 S67972.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048607 AK150430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048607 AK150830.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048607 AK152295.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762960 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138002 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048607 M96827.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048607 S67972.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048607 AK150430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048607 AK150830.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048607 AK152295.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235945 BC138872.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235945 M96827.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235945 S67972.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235945 AK150430.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235945 AK150830.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26157838 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138002 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06637214 - -
          Whole Mouse Genome G4122A A_51_P235945 BC138872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235945 M96827.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235945 S67972.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235945 AK150430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235945 AK150830.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105218 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138002 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07083793 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07392836 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07392841 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07392843 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07392845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07392844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26157827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06872983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06868858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07149557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07211462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06680921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06671530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07083793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07065158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26157856 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10581608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581606 - -
          Affymetrix HT MG-430 PM Array Plate 1448881_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10581605 - -
          Mouse Genome 430 2.0 Array 1448881_at - -
          Mouse Genome 430 2.0 Array 1448881_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15439_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15439_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800022318.mm.1 NM_017370.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022318.mm.1 NM_017370.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022318.mm.1 TC0800002905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022320.mm.1 NM_017370.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022320.mm.1 NM_017370.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022320.mm.1 TC0800002905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022321.mm.1 NM_017370.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022321.mm.1 NM_017370.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022321.mm.1 TC0800002905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022315.mm.1 NM_017370.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022315.mm.1 NM_017370.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022315.mm.1 TC0800002905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022316.mm.1 NM_017370.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022316.mm.1 NM_017370.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022316.mm.1 TC0800002905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022317.mm.1 NM_017370.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022317.mm.1 NM_017370.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022317.mm.1 TC0800002905.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022319.mm.1 NM_017370.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022319.mm.1 NM_017370.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022319.mm.1 TC0800002905.mm.1 MTA TC
          Mouse Expression Set 430 1448881_PM_at - -
          Mouse Expression Set 430 1448881_at - -
          Murine Genome U74v2 Set 96092_at - -
          Murine Genome U74v2 Set 96092_PM_at - -
          Murine Genome U74v2 Set 169605_PM_at - -
          Murine Genome U74v2 Set 169605_at - -
          Mouse Genome 430A 2.0 Array 1448881_at - -
          Mouse Genome 430A 2.0 Array 1448881_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448881_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448881_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 620526 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6350068 - -
          Illumina mouse-6 v1.0 expression beadchip 2940551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2668510 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2644764 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2668510 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2644764 - -
          mouse-6 v1.1 (Illumina) scl34345.7_19-S - -
          mouse-6 v1.1 (Illumina) scl000592.1_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 2940551 - -
          Illumina Sentrix mouse-6.v1 bead array 6350068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000592.1_8-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34345.7_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940551 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2668510 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2668509 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2944824 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2668509 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2944824 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2668510 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000592.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl34345.7_19-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline