Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517103_m1
          See other BTG4 GE Assays ›
          Gene Symbol:
          Btg4
          Gene Name:
          B cell translocation gene 4
          Gene Aliases:
          C86116, PC3B
          Chromosome Location:
          Chr.9: 51102265 - 51119700 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517103_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Btg4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019493.3 NP_062366.2 4 - 5 734 64
          GenBank mRNA AB050983.2 - 4 - 5 734 64
          AJ005120.1 - 4 - 5 607 64
          AK135849.1 - 1 - 2 113 64
          AK136041.1 - 4 - 5 721 64
          AK136124.1 - 4 - 5 722 64
          AK136149.1 - 4 - 5 724 64
          AK136185.1 - 4 - 5 726 64
          AK161379.1 - 6 - 7 813 64
          AK163407.1 - 4 - 5 722 64
          BC066811.1 - 4 - 5 708 30475849 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000613.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Btg4

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of mitotic cell cycle
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491190 AK163407.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491190 AK136149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491190 AK136185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491190 AB050983.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491190 AK136041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491190 AK163407.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491190 AK136149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491190 AK136185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491190 AB050983.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P491190 AK136041.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 559734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686022 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52000 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116375 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491190 AK163407.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491190 AK136149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491190 AK136185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491190 AB050983.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491190 AK136041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491190 AK163407.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491190 AK136149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491190 AK136185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491190 AB050983.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P491190 AK136041.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491190 AK163407.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491190 AK136149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491190 AK136185.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491190 AB050983.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491190 AK136041.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491190 AK163407.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491190 AK136149.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491190 AK136185.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491190 AB050983.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P491190 AK136041.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26507583 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06786688 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01931032 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116375 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06786671 - -
          Whole Mouse Genome G4122A A_51_P491190 AK163407.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491190 AK136149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491190 AK136185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491190 AB050983.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491190 AK136041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491190 AK163407.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491190 AK136149.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491190 AK136185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491190 AB050983.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P491190 AK136041.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120534 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116375 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06786671 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07633440 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08016836 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08016835 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07633465 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07633448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26507573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07898969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07908254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07999281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07545468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07633464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07545463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07466911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07999287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08170578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07986371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07633462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07799120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07633439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26507626 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10585269 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585274 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585272 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585271 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585270 - -
          Affymetrix HT MG-430 PM Array Plate 1426520_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10585268 - -
          Mouse Genome 430 2.0 Array 1426520_PM_at - -
          Mouse Genome 430 2.0 Array 1426520_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56057_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56057_at - -
          Affymetrix Mouse Clariom S Array PSR0900004419.mm.1 NM_019493.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004419.mm.1 TC0900000613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900004418.mm.1 NM_019493.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900004418.mm.1 TC0900000613.mm.1 MTA TC
          Mouse Expression Set 430 1426520_PM_at - -
          Mouse Expression Set 430 1426520_at - -
          Murine Genome U74v2 Set 98583_PM_at - -
          Murine Genome U74v2 Set 98583_at - -
          Mouse Genome 430A 2.0 Array 1426520_at - -
          Mouse Genome 430A 2.0 Array 1426520_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426520_at - -
          Affymetrix HT Mouse Genome 430A Array 1426520_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 735892 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4060020 - -
          Illumina mouse-6 v1.0 expression beadchip 4060020 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060020 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248124 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248124 - -
          mouse-6 v1.1 (Illumina) scl36964.9.1_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 4060020 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36964.9.1_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060020 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248124 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248124 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline