Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517434_s1
          See other CLDN9 GE Assays ›
          Gene Symbol:
          Cldn9
          Gene Name:
          claudin 9
          Gene Aliases:
          nmf329
          Chromosome Location:
          Chr.17: 23682584 - 23684026 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517434_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cldn9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020293.3 NP_064689.2 1 735 73
          GenBank mRNA BC058186.1 - 1 727 5320961 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cldn9

          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-cell junction organization
          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506961 NM_020293.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506961 BC058186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506961 NM_020293.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P506961 BC058186.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311197 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111265 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506961 NM_020293.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506961 BC058186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506961 NM_020293.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P506961 BC058186.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506961 NM_020293.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506961 BC058186.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506961 NM_020293.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P506961 BC058186.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31093043 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111265 - -
          Whole Mouse Genome G4122A A_51_P506961 NM_020293.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506961 BC058186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506961 NM_020293.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P506961 BC058186.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111936 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111265 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13540454 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13540438 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13540447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13540429 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13540443 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13540456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13540456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31093042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448313 - -
          Affymetrix HT MG-430 PM Array Plate 1450524_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439427_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10448312 - -
          Mouse Genome 430 2.0 Array 1439427_PM_at - -
          Mouse Genome 430 2.0 Array 1439427_at - -
          Mouse Genome 430 2.0 Array 1450524_PM_at - -
          Mouse Genome 430 2.0 Array 1450524_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56863_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56863_at - -
          Mouse Expression Set 430 1450524_PM_at - -
          Mouse Expression Set 430 1450524_at - -
          Mouse Expression Set 430 1439427_at - -
          Mouse Expression Set 430 1439427_PM_at - -
          Mouse Genome 430A 2.0 Array 1450524_at - -
          Mouse Genome 430A 2.0 Array 1450524_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450524_at - -
          Affymetrix HT Mouse Genome 430A Array 1450524_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439427_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439427_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 840373 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000840373 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2607331 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2607331 - -
          mouse-6 v1.1 (Illumina) scl7923.1.1_249-S - -
          Illumina Sentrix mouse-6.v1 bead array 840373 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl7923.1.1_249-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 840373 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2607331 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2607331 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline