Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517522_m1
          See other CRYBB3 GE Assays ›
          Gene Symbol:
          Crybb3
          Gene Name:
          crystallin, beta B3
          Gene Aliases:
          AI852419
          Chromosome Location:
          Chr.5: 113075839 - 113082233 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517522_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crybb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159650.1 NP_001153122.1 5 - 6 524 82
          NM_021352.3 NP_067327.1 5 - 6 594 82
          GenBank mRNA AJ272229.2 - 4 - 5 578 82
          BC031442.1 - 4 - 5 585 4501690 82
          BC058522.1 - 4 - 5 503 6824229 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003026.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crybb3

          Gene Ontology Categories:

          Process(es)

          structural constituent of eye lens

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983921 BC031442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983921 BC058522.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983921 CJ046471.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983921 AK053552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983921 NM_001159650.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519486 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43391 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 19027 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106577 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983921 BC031442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983921 BC058522.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983921 CJ046471.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983921 AK053552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983921 NM_001159650.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P286511 BC031442.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P286511 BC058522.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P286511 CJ046471.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P286511 AK053552.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P286511 NM_001159650.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23996024 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106577 - -
          Whole Mouse Genome G4122A A_52_P286511 BC031442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P286511 BC058522.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P286511 CJ046471.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P286511 AK053552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P286511 NM_001159650.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127863 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106577 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04617303 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04551716 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04551715 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04471868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04551722 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04471869 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04551720 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04551725 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04471870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23995992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23996067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04130273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04617302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04516057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04837689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04130276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04306442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04617305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04130267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04551720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04471870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04412326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04130270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04516056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04959181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04837692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23996020 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532661 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532663 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532668 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532664 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532662 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532667 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532666 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532665 - -
          Affymetrix HT MG-430 PM Array Plate 1419005_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10532660 - -
          Mouse Genome 430 2.0 Array 1419005_at - -
          Mouse Genome 430 2.0 Array 1419005_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12962_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12962_at - -
          Affymetrix Mouse Clariom S Array PSR0500026907.mm.1 TC0500003026.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026907.mm.1 NM_001159650.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026907.mm.1 NM_001159650.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026907.mm.1 NM_021352.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026907.mm.1 NM_021352.3 Refseq
          Mouse Expression Set 430 1419005_PM_at - -
          Mouse Expression Set 430 1419005_at - -
          Mouse Genome 430A 2.0 Array 1419005_PM_at - -
          Mouse Genome 430A 2.0 Array 1419005_at - -
          Affymetrix HT Mouse Genome 430A Array 1419005_at - -
          Affymetrix HT Mouse Genome 430A Array 1419005_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 354201 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6100082 - -
          Illumina mouse-6 v1.0 expression beadchip 102640402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006100082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720107 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2720107 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251317 - -
          mouse-6 v1.1 (Illumina) scl26202.7.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 6100082 - -
          Illumina Sentrix mouse-6.v1 bead array 102640402 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26202.7.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100082 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102640402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2720107 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2720107 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251317 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline