Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517529_m1
          See other CHRNA6 GE Assays ›
          Gene Symbol:
          Chrna6
          Gene Name:
          cholinergic receptor, nicotinic, alpha polypeptide 6
          Gene Aliases:
          Acra6, Nica6
          Chromosome Location:
          Chr.8: 27403212 - 27413944 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517529_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chrna6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021369.2 NP_067344.2 1 - 2 327 85
          GenBank mRNA AJ245706.1 - 1 - 2 165 85
          AK149262.1 - 1 - 2 268 85
          AY574265.1 - 1 - 2 85 85
          BC031985.1 - 1 - 2 327 4501558 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001959.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chrna6

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          regulation of dopamine secretion
          positive regulation of dopamine secretion
          protein heterooligomerization
          membrane depolarization
          cation transmembrane transport
          acetylcholine-activated cation-selective channel activity
          ion channel activity
          extracellular ligand-gated ion channel activity
          protein binding
          drug binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041065 BC031985.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041065 AK149262.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041065 NM_021369.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494438 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179978 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138736 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041065 BC031985.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041065 AK149262.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041065 NM_021369.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P499653 BC031985.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P499653 AK149262.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P499653 AY574265.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P499653 AJ245706.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P499653 NM_021369.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25738156 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138736 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179978 - -
          Whole Mouse Genome G4122A A_51_P499653 BC031985.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499653 AK149262.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499653 AY574265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499653 AJ245706.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499653 NM_021369.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129660 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140820 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179978 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138736 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06898819 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06898812 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06898817 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06898813 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06892137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25738165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06853125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06910151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07136594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07136422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07203660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06853114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06892141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07136587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06892140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25738153 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10577985 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577983 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577984 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577982 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577986 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577987 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577989 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577988 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577981 - -
          Affymetrix HT MG-430 PM Array Plate 1450427_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1450426_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10577980 - -
          Mouse Genome 430 2.0 Array 1450426_at - -
          Mouse Genome 430 2.0 Array 1450426_PM_at - -
          Mouse Genome 430 2.0 Array 1450427_at - -
          Mouse Genome 430 2.0 Array 1450427_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11440_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11440_at - -
          Affymetrix Mouse Clariom S Array PSR0800015561.mm.1 NM_021369.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015561.mm.1 NM_021369.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015561.mm.1 TC0800001959.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800015562.mm.1 NM_021369.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015562.mm.1 NM_021369.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800015562.mm.1 TC0800001959.mm.1 MTA TC
          Mouse Expression Set 430 1450427_PM_at - -
          Mouse Expression Set 430 1450426_PM_at - -
          Mouse Expression Set 430 1450426_at - -
          Mouse Expression Set 430 1450427_at - -
          Murine Genome U74v2 Set 138177_PM_at - -
          Murine Genome U74v2 Set 138177_at - -
          Mouse Genome 430A 2.0 Array 1450426_at - -
          Mouse Genome 430A 2.0 Array 1450427_at - -
          Mouse Genome 430A 2.0 Array 1450427_PM_at - -
          Mouse Genome 430A 2.0 Array 1450426_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450427_at - -
          Affymetrix HT Mouse Genome 430A Array 1450426_at - -
          Affymetrix HT Mouse Genome 430A Array 1450427_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450426_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 395875 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5340092 - -
          Illumina mouse-6 v1.0 expression beadchip 5340092 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005340092 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2732438 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2732438 - -
          mouse-6 v1.1 (Illumina) scl34973.6.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 5340092 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34973.6.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5340092 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2732437 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2732438 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2732437 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2732438 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline