Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517732_m1
          See other DAP3 GE Assays ›
          Gene Symbol:
          Dap3
          Gene Name:
          death associated protein 3
          Gene Aliases:
          4921514D13Rik, DAP-3, MRP-S29, S29mt
          Chromosome Location:
          Chr.3: 88920803 - 88951266 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517732_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dap3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164533.1 NP_001158005.1 2 - 3 470 110
          NM_022994.3 NP_075370.2 2 - 3 470 110
          GenBank mRNA AJ250375.1 - 2 - 3 214 110
          AK014280.1 - 2 - 3 140 110
          AK014889.1 - 2 - 3 140 110
          AK032275.1 - 2 - 3 142 110
          AK088960.1 - 2 - 3 140 110
          BC019566.1 - 2 - 3 470 4235517 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002329.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dap3

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          apoptotic mitochondrial changes
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22316451 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22316450 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975743 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975742 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242695 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 525363 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 958856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245864 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444006 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384691 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807093 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493088 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 119897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 73391 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975743 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975742 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22316455 - -
          Whole Mouse Genome G4122A A_52_P292882 - -
          Whole Mouse Genome G4122A A_51_P235580 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129434 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02408434 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02408435 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22316450 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22316451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22316470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02565657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03058263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02585906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02487603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02963980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22316459 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499457 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499458 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499462 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499464 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499467 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499463 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499469 - -
          Affymetrix HT MG-430 PM Array Plate 1450848_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10499454 - -
          Mouse Genome 430 2.0 Array 1450848_at - -
          Mouse Genome 430 2.0 Array 1450848_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 65111_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 65111_at - -
          Affymetrix Mouse Clariom S Array PSR0300017791.mm.1 NM_001164533.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017791.mm.1 NM_001164533.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017791.mm.1 NM_022994.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017791.mm.1 NM_022994.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017791.mm.1 TC0300002329.mm.1 MTA TC
          Murine Genome U74v2 Set 94524_at - -
          Murine Genome U74v2 Set 94524_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450848_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450848_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 568479 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1660528 - -
          Illumina mouse-6 v1.0 expression beadchip 2120039 - -
          Illumina mouse-6 v1.0 expression beadchip 5420593 - -
          Illumina mouse-6 v1.0 expression beadchip 1660528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005420593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120039 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2722542 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620078 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646230 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620078 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2722542 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2646230 - -
          mouse-6 v1.1 (Illumina) scl0001977.1_31-S - -
          mouse-6 v1.1 (Illumina) scl065111.1_137-S - -
          mouse-6 v1.1 (Illumina) scl0065111.1_63-S - -
          Illumina Sentrix mouse-6.v1 bead array 1660528 - -
          Illumina Sentrix mouse-6.v1 bead array 2120039 - -
          Illumina Sentrix mouse-6.v1 bead array 5420593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0065111.1_63-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl065111.1_137-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001977.1_31-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660528 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120039 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5420593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2646230 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2620078 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2953910 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2722542 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620078 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2646230 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2953910 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2722542 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0065111.1_63-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl065111.1_137-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001977.1_31-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline