Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517765_g1
          See other GNGT2 GE Assays ›
          Gene Symbol:
          Gngt2
          Gene Name:
          guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 2
          Gene Aliases:
          AV096488
          Chromosome Location:
          Chr.11: 95842219 - 95845734 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517765_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gngt2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001038664.2 NP_001033753.2 3 - 4 320 82
          NM_001284393.1 NP_001271322.1 3 - 4 324 82
          NM_001284397.1 NP_001271326.1 3 - 4 321 82
          NM_023121.2 NP_075610.1 3 - 4 305 82
          GenBank mRNA AA028742.1 - 3 - 4 275 82
          AA423571.1 - 3 - 4 194 82
          AI159118.1 - 3 - 4 262 82
          AK010554.1 - 2 - 3 1334 82
          BC061005.1 - 3 - 4 270 30295552 82
          BE847751.1 - 3 - 4 310 82
          BE994683.1 - 1 - 2 82
          BF470980.1 - 2 - 3 134 82

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gngt2

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          metabolic process
          signal transducer activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151986 BC061005.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151986 BE994683.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151986 BF470980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151986 NM_001038664.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151986 NM_023121.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333234 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431117 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151986 BC061005.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151986 BE994683.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151986 BF470980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151986 NM_001038664.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151986 NM_023121.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28041156 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02401659 - -
          Whole Mouse Genome G4122A A_52_P44824 AK010554.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112527 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08982790 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08982799 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08982796 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08982771 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08982797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28041159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09272994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08982780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09153026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09272991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28041201 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10380572 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380576 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380573 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380577 - -
          Affymetrix HT MG-430 PM Array Plate 1428733_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10380571 - -
          Mouse Genome 430 2.0 Array 1428733_at - -
          Mouse Genome 430 2.0 Array 1428733_PM_at - -
          Mouse Expression Set 430 1428733_at - -
          Mouse Expression Set 430 1428733_PM_at - -
          Murine Genome U74v2 Set 160893_at - -
          Murine Genome U74v2 Set 160893_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428733_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428733_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 846124 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 540176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540176 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246056 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246056 - -
          mouse-6 v1.1 (Illumina) scl41002.6.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 540176 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41002.6.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540176 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246056 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3154581 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3154581 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246056 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0