Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517807_m1
          See other HIST1H2BC GE Assays ›
          Gene Symbol:
          Hist1h2bc
          Gene Name:
          histone cluster 1, H2bc
          Gene Aliases:
          2610022J01Rik, H2bfs, R74621
          Chromosome Location:
          Chr.13: 23684199 - 23692480 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517807_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist1h2bc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023422.3 NP_075911.2 1 - 2 424 94
          GenBank mRNA AK005191.1 - 1 - 2 420 94
          AK005407.1 - 1 - 2 419 94
          AK011516.1 - 1 - 2 426 94
          AK208955.1 - 1 - 2 59 94
          BC082774.1 - 1 - 2 407 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000255.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hist1h2bc

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          nucleosome assembly
          antibacterial humoral response
          defense response to Gram-positive bacterium
          molecular_function
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882834 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109122 BC019673.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109122 BC082774.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109122 NM_023422.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109122 AK005191.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109122 AK005407.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 414298 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32870 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667870 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313727 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282128 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 85636 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109122 BC019673.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109122 BC082774.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109122 NM_023422.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109122 AK005191.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109122 AK005407.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P462311 AK005191.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882859 - -
          Whole Mouse Genome G4122A A_51_P462311 AK005191.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110661 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10491561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10491534 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10491541 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10491549 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10491553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10576839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10491531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10491547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10631703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10491543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10576846 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403979 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408211 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403960 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403981 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408091 - -
          Affymetrix HT MG-430 PM Array Plate 1418072_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10408087 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408223 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403959 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404053 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408210 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408070 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403980 - -
          Mouse Genome 430 2.0 Array 1452540_a_at - -
          Mouse Genome 430 2.0 Array 1425398_at - -
          Mouse Genome 430 2.0 Array 1418072_PM_at - -
          Mouse Genome 430 2.0 Array 1452540_PM_a_at - -
          Mouse Genome 430 2.0 Array 1425398_PM_at - -
          Mouse Genome 430 2.0 Array 1418072_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68024_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68024_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300001428.mm.1 NM_023422.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001428.mm.1 NM_023422.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001428.mm.1 TC1300000255.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001429.mm.1 NM_023422.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001429.mm.1 NM_023422.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001429.mm.1 TC1300000255.mm.1 MTA TC
          Mouse Expression Set 430 1418072_at - -
          Mouse Expression Set 430 1425398_at - -
          Mouse Expression Set 430 1425398_PM_at - -
          Mouse Expression Set 430 1452540_PM_a_at - -
          Mouse Expression Set 430 1418072_PM_at - -
          Mouse Expression Set 430 1452540_a_at - -
          Murine Genome U74v2 Set 93833_PM_s_at - -
          Murine Genome U74v2 Set 101654_PM_at - -
          Murine Genome U74v2 Set 93833_s_at - -
          Murine Genome U74v2 Set 101654_at - -
          Mouse Genome 430A 2.0 Array 1418072_PM_at - -
          Mouse Genome 430A 2.0 Array 1452540_a_at - -
          Mouse Genome 430A 2.0 Array 1418072_at - -
          Mouse Genome 430A 2.0 Array 1425398_at - -
          Mouse Genome 430A 2.0 Array 1452540_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1425398_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 929520 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6520315 - -
          Illumina mouse-6 v1.0 expression beadchip 2570156 - -
          Illumina mouse-6 v1.0 expression beadchip 6520315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520315 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242399 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2677231 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2677231 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242399 - -
          mouse-6 v1.1 (Illumina) scl068024.2_210-S - -
          mouse-6 v1.1 (Illumina) scl44991.2.1_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 2570156 - -
          Illumina Sentrix mouse-6.v1 bead array 6520315 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44991.2.1_21-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068024.2_210-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520315 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570156 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242399 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2677231 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242399 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2677231 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl44991.2.1_21-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-snt5b:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline