Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517828_m1
          See other CRYBB1 GE Assays ›
          Gene Symbol:
          Crybb1
          Gene Name:
          crystallin, beta B1
          Gene Aliases:
          3110006K12Rik, BB1CRY
          Chromosome Location:
          Chr.5: 112254477 - 112269582 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517828_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crybb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023695.2 NP_076184.1 5 - 6 608 58
          GenBank mRNA AF106853.1 - 5 - 6 608 58
          AK014012.1 - 5 - 6 610 58
          AK053869.1 - 5 - 6 609 58
          BC029012.1 - 4 - 5 592 5358714 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001178.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crybb1

          Gene Ontology Categories:

          Process(es)

          structural constituent of eye lens
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136730 AF106853.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136730 AK014012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136730 NM_023695.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136730 AK053869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136730 BC029012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334063 AF106853.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334063 AK014012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334063 NM_023695.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334063 AK053869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334063 BC029012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334063 AF106853.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334063 AK014012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334063 NM_023695.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334063 AK053869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P334063 BC029012.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464941 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134068 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63339 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169487 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136730 AF106853.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136730 AK014012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136730 NM_023695.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136730 AK053869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136730 BC029012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334063 AF106853.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334063 AK014012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334063 NM_023695.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334063 AK053869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334063 BC029012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334063 AF106853.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334063 AK014012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334063 NM_023695.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334063 AK053869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P334063 BC029012.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334063 AF106853.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334063 AK014012.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334063 NM_023695.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334063 AK053869.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334063 BC029012.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334063 AF106853.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334063 AK014012.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334063 NM_023695.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334063 AK053869.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334063 BC029012.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169487 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05698076 - -
          Whole Mouse Genome G4122A A_51_P334063 AF106853.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334063 AK014012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334063 NM_023695.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334063 AK053869.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334063 BC029012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334063 AF106853.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334063 AK014012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334063 NM_023695.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334063 AK053869.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334063 BC029012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643798 AF106853.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643798 AK014012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643798 NM_023695.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643798 AK053869.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643798 BC029012.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109644 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105812 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04821914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04821913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04731010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04731011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04465732 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10524339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524344 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524341 - -
          Affymetrix HT MG-430 PM Array Plate 1418306_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10524338 - -
          Mouse Genome 430 2.0 Array 1418306_PM_at - -
          Mouse Genome 430 2.0 Array 1418306_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12960_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12960_at - -
          Affymetrix Mouse Clariom S Array PSR0500010056.mm.1 TC0500001178.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010056.mm.1 NM_023695.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010057.mm.1 TC0500001178.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010057.mm.1 NM_023695.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500010058.mm.1 TC0500001178.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500010058.mm.1 NM_023695.3 Refseq
          Mouse Expression Set 430 1418306_PM_at - -
          Mouse Expression Set 430 1418306_at - -
          Mouse Genome 430A 2.0 Array 1418306_PM_at - -
          Mouse Genome 430A 2.0 Array 1418306_at - -
          Affymetrix HT Mouse Genome 430A Array 1418306_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418306_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 911316 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3170095 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003170095 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2702122 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2702122 - -
          mouse-6 v1.1 (Illumina) scl27413.2.1_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 3170095 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27413.2.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170095 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2702122 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2702122 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27413.2.1_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline