Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517851_m1
          See other PRL5A1 GE Assays ›
          Gene Symbol:
          Prl5a1
          Gene Name:
          prolactin family 5, subfamily a, member 1
          Gene Aliases:
          1600013P04Rik, D13Wsu14e, PLP-L, Prlpl
          Chromosome Location:
          Chr.13: 28142484 - 28151595 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517851_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prl5a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023746.4 NP_076235.1 1 - 2 95 75
          GenBank mRNA AF226611.1 - 1 - 2 42 75
          AK005447.1 - 1 - 2 96 75
          BC051672.1 - 1 - 2 82 30017843 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000309.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prl5a1

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320606 AF226611.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320606 AK005447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320606 BC051672.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320606 NM_023746.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320606 AF226611.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320606 AK005447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320606 BC051672.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320606 NM_023746.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181933 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292337 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114964 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121080 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320606 AF226611.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320606 AK005447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320606 BC051672.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320606 NM_023746.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320606 AF226611.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320606 AK005447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320606 BC051672.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320606 NM_023746.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28904689 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121080 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02652681 - -
          Whole Mouse Genome G4122A A_51_P320606 AF226611.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320606 AK005447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320606 BC051672.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320606 NM_023746.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320606 AF226611.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320606 AK005447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320606 BC051672.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320606 NM_023746.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120458 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138620 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121080 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11030315 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10688087 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10688088 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10688081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10688090 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10688085 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11030316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28904732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10688088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11073091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10688081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28904730 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404351 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404348 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404349 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404353 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404350 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404352 - -
          Affymetrix HT MG-430 PM Array Plate 1422657_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10404347 - -
          Mouse Genome 430 2.0 Array 1422657_at - -
          Mouse Genome 430 2.0 Array 1422657_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 28078_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 28078_at - -
          Affymetrix Mouse Clariom S Array PSR1300001918.mm.1 NM_023746.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001918.mm.1 NM_023746.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001918.mm.1 TC1300000309.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001919.mm.1 NM_023746.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001919.mm.1 NM_023746.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001919.mm.1 TC1300000309.mm.1 MTA TC
          Mouse Expression Set 430 1422657_PM_at - -
          Mouse Expression Set 430 1422657_at - -
          Murine Genome U74v2 Set 168707_PM_i_at - -
          Murine Genome U74v2 Set 168707_i_at - -
          Mouse Genome 430A 2.0 Array 1422657_PM_at - -
          Mouse Genome 430A 2.0 Array 1422657_at - -
          Affymetrix HT Mouse Genome 430A Array 1422657_at - -
          Affymetrix HT Mouse Genome 430A Array 1422657_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 655380 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4480528 - -
          Illumina mouse-6 v1.0 expression beadchip 4480528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004480528 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2676207 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2676207 - -
          mouse-6 v1.1 (Illumina) scl44960.4.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 4480528 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44960.4.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4480528 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2896505 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2676207 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2896505 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2676207 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline