Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517985_m1
          See other ZWINT GE Assays ›
          Gene Symbol:
          Zwint
          Gene Name:
          ZW10 interactor
          Gene Aliases:
          2010007E07Rik, 2600001N01Rik, D10Ertd749e, Zwint-1
          Chromosome Location:
          Chr.10: 72654846 - 72674964 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517985_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zwint
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001293683.1 NP_001280612.1 1 - 2 132 102
          NM_001293684.1 NP_001280613.1 1 - 2 132 102
          NM_025635.4 NP_079911.1 1 - 2 132 102
          GenBank mRNA AK004305.1 - 1 - 2 84 102
          AK008144.1 - 1 - 2 82 102
          AK013556.1 - 1 - 2 67 102
          AK075977.1 - 1 - 2 78 102
          AK082215.1 - 1 - 2 82 102
          AK134380.1 - 1 - 2 84 102
          BC027100.1 - 1 - 2 89 5359154 102
          BC034870.1 - 1 - 2 61 1382192 102
          BY240779.1 - 1 - 2 133 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000693.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zwint

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          cell cycle
          mitotic nuclear division
          mitotic cell cycle checkpoint
          cell division
          establishment of localization in cell
          protein N-terminus binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000004823.mm.1 NM_025635.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004823.mm.1 TC1000000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000004831.mm.1 NM_025635.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004831.mm.1 TC1000000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000004825.mm.1 NM_025635.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004825.mm.1 TC1000000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000004832.mm.1 NM_025635.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004832.mm.1 TC1000000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000004827.mm.1 NM_025635.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004827.mm.1 TC1000000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000004829.mm.1 NM_025635.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000004829.mm.1 TC1000000693.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 680985 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline