Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517999_m1
          See other KLHL10 GE Assays ›
          Gene Symbol:
          Klhl10
          Gene Name:
          kelch-like 10
          Gene Aliases:
          4921517C11Rik
          Chromosome Location:
          Chr.11: 100441924 - 100457027 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517999_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klhl10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025727.3 NP_080003.1 3 - 4 1411 76
          GenBank mRNA AK006288.1 - 3 - 4 1398 76
          AK014906.1 - 3 - 4 1412 76
          AK016593.1 - 1 - 2 477 76
          AK077202.1 - 3 - 4 1413 76
          AY495337.1 - 3 - 4 1412 76
          BC139267.1 - 3 - 4 1387 8862289 76
          BC139268.1 - 3 - 4 1387 8862290 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001625.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klhl10

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          spermatogenesis
          spermatid development
          male gonad development
          fertilization
          cell differentiation
          male genitalia morphogenesis
          homeostasis of number of cells within a tissue
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036480 BC139267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036480 BC139268.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036480 AY495337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036480 AK014906.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036480 NM_025727.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377005 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432508 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 90895 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 533 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108319 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176705 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036480 BC139267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036480 BC139268.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036480 AY495337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036480 AK014906.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036480 NM_025727.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P137254 BC139267.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P137254 BC139268.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P137254 AK077202.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P137254 AY495337.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P137254 AK006288.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28068137 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176705 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108319 - -
          Whole Mouse Genome G4122A A_51_P137254 BC139267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137254 BC139268.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137254 AK077202.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137254 AY495337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137254 AK006288.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122483 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176705 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28068150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09010196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09286888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09413776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09010205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09314597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09413771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09413777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09252322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09252326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09252329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09314591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09252339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28068142 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381134 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381138 - -
          Affymetrix HT MG-430 PM Array Plate 1423497_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10381133 - -
          Mouse Genome 430 2.0 Array 1423497_at - -
          Mouse Genome 430 2.0 Array 1423497_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66720_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66720_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100015047.mm.1 NM_025727.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015047.mm.1 NM_025727.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015047.mm.1 TC1100001625.mm.1 MTA TC
          Mouse Expression Set 430 1423497_PM_at - -
          Mouse Expression Set 430 1423497_at - -
          Mouse Genome 430A 2.0 Array 1423497_at - -
          Mouse Genome 430A 2.0 Array 1423497_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423497_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423497_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 352384 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 780139 - -
          Illumina mouse-6 v1.0 expression beadchip 780139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2724860 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2724860 - -
          mouse-6 v1.1 (Illumina) scl40909.5.1_222-S - -
          Illumina Sentrix mouse-6.v1 bead array 780139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40909.5.1_222-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2921031 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2921031 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline