| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22384844 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22384842 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22384843 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P180408 |
BC023382.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P180408 |
BC037062.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P180408 |
AK153513.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P180408 |
AK166967.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P180408 |
NM_026602.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P180408 |
BC023382.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P180408 |
BC037062.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P180408 |
AK153513.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P180408 |
AK166967.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P180408 |
NM_026602.3 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
274351 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
64981 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
333594 |
NM_001104634.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
333594 |
NM_001104635.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
333594 |
NM_001104637.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
333594 |
XM_001003047.2 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
333594 |
XM_621729.4 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
333594 |
XM_887513.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
333594 |
XM_988849.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
138597 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P160107 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P138092 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P180408 |
BC023382.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P180408 |
BC037062.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P180408 |
AK153513.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P180408 |
AK166967.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P180408 |
NM_026602.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P180408 |
BC023382.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P180408 |
BC037062.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P180408 |
AK153513.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P180408 |
AK166967.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P180408 |
NM_026602.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P29513 |
BC013627.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P29513 |
BC023382.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P29513 |
BC037062.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P29513 |
AK153513.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P29513 |
AK166967.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P180408 |
BC023382.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P180408 |
BC037062.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P180408 |
AK153513.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P180408 |
AK166967.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P180408 |
NM_026602.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P180408 |
BC023382.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P180408 |
BC037062.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P180408 |
AK153513.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P180408 |
AK166967.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P180408 |
NM_026602.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P467990 |
AK018786.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384842 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384854 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384853 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384846 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384847 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384835 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384845 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384848 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384844 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384850 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384857 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384841 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384838 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22384852 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P160107 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P138092 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P180408 |
BC023382.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P180408 |
BC037062.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P180408 |
AK153513.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P180408 |
AK166967.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P180408 |
NM_026602.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P180408 |
BC023382.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P180408 |
BC037062.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P180408 |
AK153513.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P180408 |
AK166967.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P180408 |
NM_026602.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P29513 |
BC013627.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P29513 |
BC023382.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P29513 |
BC037062.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P29513 |
AK153513.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P29513 |
AK166967.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P467990 |
AK018786.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P119240 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P138092 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P160107 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02862519 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03041943 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03041945 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02862522 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02862525 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22384842 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22384844 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22384843 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384842 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384915 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384887 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384859 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384841 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384918 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384909 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384908 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384889 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384925 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384928 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384888 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384852 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384893 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384846 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384929 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384905 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384896 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384878 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384901 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384837 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384895 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384863 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384853 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384897 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384875 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384844 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384862 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384877 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384845 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384922 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384873 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384881 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384880 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384906 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384882 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384927 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384835 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384869 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384855 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384903 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384867 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384911 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384884 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384891 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384857 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22384848 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384842 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384850 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384893 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03173236 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384857 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02738499 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384837 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384889 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384884 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384878 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384853 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384841 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02738479 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384885 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384896 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384873 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384880 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384866 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02738485 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384848 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384865 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384869 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384835 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384859 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02738480 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384891 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384852 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384862 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384844 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384888 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384839 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384881 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384877 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02372649 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384863 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384897 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02862522 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22384847 |
- |
- |