Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00518388_mH
          See other CEACAM13 GE Assays ›
          Gene Symbol:
          Ceacam13
          Gene Name:
          carcinoembryonic antigen-related cell adhesion molecule 13
          Gene Aliases:
          1600012K03Rik, 1600025J19Rik
          Chromosome Location:
          Chr.7: 18009889 - 18019220 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00518388_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ceacam13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027210.3 NP_081486.1 1 - 2 144 102
          NM_028171.2 NP_082447.1 1 - 2 144 102
          GenBank mRNA AK005430.1 - 1 - 2 135 102
          AK005537.1 - 1 - 2 145 102
          AK146045.1 - 1 - 2 145 102
          BC099430.1 - 1 - 2 131 30787167 102
          BY705748.1 - 1 - 2 145 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000262.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ceacam13

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000117 NM_028171.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000117 AK005430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000117 AK005537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000117 AK146045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000117 BC099430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179299 BC099430.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 745833 AK006881.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 560459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666830 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179299 BC099430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000117 NM_028171.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000117 AK005430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000117 AK005537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000117 AK146045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000117 BC099430.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24987183 - -
          Whole Mouse Genome G4122A A_51_P429743 AK005430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429743 AK146045.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429743 BC099430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P429743 NM_027210.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P160155 NM_028171.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P160155 AK005430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P160155 AK005537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P160155 AK146045.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P160155 BC099430.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104152 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975120 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975124 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06020402 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24987183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05975113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06020426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06020429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06020402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06020434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06553340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05975104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06378218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06020391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06020436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06161989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24987190 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550473 - -
          Affymetrix HT MG-430 PM Array Plate 1431647_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550470 - -
          Mouse Genome 430 2.0 Array 1431647_PM_a_at - -
          Mouse Genome 430 2.0 Array 1431647_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69785_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69785_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700001937.mm.1 TC0700000262.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001937.mm.1 NM_028171.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001937.mm.1 NM_028171.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001936.mm.1 TC0700000262.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001936.mm.1 NM_027210.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001936.mm.1 NM_027210.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001936.mm.1 NM_028171.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001936.mm.1 NM_028171.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001935.mm.1 TC0700000262.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001935.mm.1 NM_027210.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001935.mm.1 NM_027210.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001935.mm.1 NM_028171.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001935.mm.1 NM_028171.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001934.mm.1 TC0700000262.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001934.mm.1 NM_027210.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001934.mm.1 NM_027210.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001934.mm.1 NM_028171.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001934.mm.1 NM_028171.2 Refseq
          Murine Genome U74v2 Set 165879_at - -
          Murine Genome U74v2 Set 165879_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431647_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1431647_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 407790 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220451 - -
          MouseWG-6 v2 Expression BeadChip 4480129 - -
          Illumina mouse-6 v1.0 expression beadchip 5220451 - -
          Illumina mouse-6 v1.0 expression beadchip 4480129 - -
          Illumina mouse-6 v1.0 expression beadchip 60452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000060452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004480129 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692200 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2642321 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2592257 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692200 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2642321 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2592257 - -
          mouse-6 v1.1 (Illumina) scl00086.1_13-S - -
          mouse-6 v1.1 (Illumina) scl000229.1_6-S - -
          mouse-6 v1.1 (Illumina) scl33028.3.224_37-S - -
          Illumina Sentrix mouse-6.v1 bead array 60452 - -
          Illumina Sentrix mouse-6.v1 bead array 4480129 - -
          Illumina Sentrix mouse-6.v1 bead array 5220451 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33028.3.224_37-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000229.1_6-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00086.1_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 60452 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220451 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4480129 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2692200 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2642321 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2592257 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2592257 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2642321 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2692200 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000229.1_6-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline