Search
Search

Ppp6r3| Gene Symbol: Ppp6r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164159.1 | NP_001157631.1 | 12 - 13 | 1575 | 55 | |
| NM_028999.1 | NP_083275.1 | 13 - 14 | 1723 | 55 | ||
| NM_029456.2 | NP_083732.1 | 13 - 14 | 1723 | 55 | ||
| GenBank mRNA | AK018652.1 | - | 12 - 13 | 1564 | 55 | |
| AK032179.1 | - | 4 - 5 | 494 | 55 | ||
| AK035236.1 | - | 13 - 14 | 1703 | 55 | ||
| AK088526.1 | - | 12 - 13 | 1543 | 55 | ||
| BC008529.1 | - | 13 - 14 | 1706 | 3601021 | 55 | |
| BU612294.1 | - | 1 - 2 | 117 | 55 | ||
| CF727136.1 | - | 3 - 4 | 240 | 55 | ||
Set Membership: |
|