Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00518630_m1
          See other KEG1 GE Assays ›
          Gene Symbol:
          Keg1
          Gene Name:
          kidney expressed gene 1
          Gene Aliases:
          0610008P16Rik, AI316519, GS4059
          Chromosome Location:
          Chr.19: 12695790 - 12719902 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00518630_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Keg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029550.4 NP_083826.1 5 - 6 548 100
          GenBank mRNA AB028071.1 - 5 - 6 525 100
          AK002360.1 - 5 - 6 546 100
          AK165515.1 - 5 - 6 546 100
          BC010803.1 - 5 - 6 538 4238654 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000254.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Keg1

          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Process(es)

          transferase activity
          transferase activity, transferring acyl groups
          glycine N-acyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653825 AK165515.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653825 BC010803.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653825 AI099383.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653825 AK002360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653825 AB028071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653825 AK165515.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653825 BC010803.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653825 AI099383.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653825 AK002360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653825 AB028071.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 114884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88130 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 525135 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843145 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28214 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319552 BC115976.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319552 NM_008023.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319552 X92591.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263296 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40685 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128384 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109382 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107758 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653825 AK165515.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653825 BC010803.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653825 AI099383.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653825 AK002360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653825 AB028071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653825 AK165515.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653825 BC010803.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653825 AI099383.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653825 AK002360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653825 AB028071.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31961960 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09109099 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09109112 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109382 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03552226 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107758 - -
          Whole Mouse Genome G4122A A_51_P492153 AK165515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492153 BC010803.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492153 AK002360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492153 AB028071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492153 NM_029550.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653825 AK165515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653825 BC010803.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653825 AI099383.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653825 AK002360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653825 AB028071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653825 AK165515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653825 BC010803.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653825 AI099383.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653825 AK002360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653825 AB028071.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110613 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107758 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109382 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198299 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14469612 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198302 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14198295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31961978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14158608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14469612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31961957 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461764 - -
          Affymetrix HT MG-430 PM Array Plate 1416833_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461758 - -
          Mouse Genome 430 2.0 Array 1444032_PM_at - -
          Mouse Genome 430 2.0 Array 1416833_PM_at - -
          Mouse Genome 430 2.0 Array 1416833_at - -
          Mouse Genome 430 2.0 Array 1444032_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64697_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64697_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900002991.mm.1 NM_029550.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002991.mm.1 NM_029550.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002991.mm.1 TC1900000254.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002993.mm.1 NM_029550.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002993.mm.1 NM_029550.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002993.mm.1 TC1900000254.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002994.mm.1 NM_029550.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002994.mm.1 NM_029550.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002994.mm.1 TC1900000254.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002992.mm.1 NM_029550.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002992.mm.1 NM_029550.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002992.mm.1 TC1900000254.mm.1 MTA TC
          Mouse Expression Set 430 1416833_PM_at - -
          Mouse Expression Set 430 1416833_at - -
          Murine Genome U74v2 Set 96938_at - -
          Murine Genome U74v2 Set 96938_PM_at - -
          Mouse Genome 430A 2.0 Array 1416833_PM_at - -
          Mouse Genome 430A 2.0 Array 1416833_at - -
          Affymetrix HT Mouse Genome 430A Array 1416833_at - -
          Affymetrix HT Mouse Genome 430A Array 1416833_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 889528 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6940603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940603 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2745124 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2745124 - -
          mouse-6 v1.1 (Illumina) scl53337.7_252-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53337.7_252-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2860915 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2860915 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline