Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00518698_m1
          See other CREB3L4 GE Assays ›
          Gene Symbol:
          Creb3l4
          Gene Name:
          cAMP responsive element binding protein 3-like 4
          Gene Aliases:
          1700012K17Rik, 5330432F22Rik, AIBZIP, AV040530, AV258827, CREB4, JAL, TISP40, acre1
          Chromosome Location:
          Chr.3: 90237500 - 90243524 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00518698_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Creb3l4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AB047647.1 - 2 - 3 301 116
          AB052779.1 - 4 - 5 558 116
          AB182651.1 - 4 - 5 676 116
          AB182652.1 - 2 - 3 301 116
          AF287260.1 - 2 - 3 439 116
          AK005918.1 - 2 - 3 458 116
          AK019928.1 - 4 - 5 676 116
          AK088919.1 - 4 - 5 575 116
          AY049978.1 - 4 - 5 597 116
          BC022605.1 - 4 - 5 532 4164994 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002367.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324141 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324140 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497882 BC022605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497882 AK019928.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497882 AK088919.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497882 AB052779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497882 AB182651.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497882 BC022605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497882 AK019928.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497882 AK088919.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497882 AB052779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P497882 AB182651.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653351 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25437 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152835 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497882 BC022605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497882 AK019928.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497882 AK088919.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497882 AB052779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497882 AB182651.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497882 BC022605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497882 AK019928.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497882 AK088919.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497882 AB052779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P497882 AB182651.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497882 BC022605.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497882 AK019928.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497882 AK088919.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497882 AB052779.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497882 AB182651.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497882 BC022605.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497882 AK019928.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497882 AK088919.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497882 AB052779.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P497882 AB182651.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22324110 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152835 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04985938 - -
          Whole Mouse Genome G4122A A_51_P497882 BC022605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497882 AK019928.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497882 AK088919.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497882 AB052779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497882 AB182651.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497882 BC022605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497882 AK019928.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497882 AK088919.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497882 AB052779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497882 AB182651.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109665 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152835 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03107461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02786633 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02989976 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03107457 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02989974 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03107462 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03107450 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03107472 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03107460 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324140 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22324141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22324121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02823583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02823570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03107462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03107457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02823584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02823568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02823586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02453456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02823588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02823577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02823579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02838902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03064094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03107478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02786632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22324132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03107460 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499752 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499765 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499754 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499762 - -
          Affymetrix HT MG-430 PM Array Plate 1424218_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10499751 - -
          Mouse Genome 430 2.0 Array 1424218_PM_a_at - -
          Mouse Genome 430 2.0 Array 1424218_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78284_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78284_at - -
          Affymetrix Mouse Clariom S Array PSR0300018361.mm.1 NM_030080.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018361.mm.1 TC0300002367.mm.1 MTA TC
          Mouse Expression Set 430 1424218_a_at - -
          Mouse Expression Set 430 1424218_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1424218_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1424218_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424218_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424218_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 715401 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1980736 - -
          Illumina mouse-6 v1.0 expression beadchip 1980736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980736 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213549 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213549 - -
          mouse-6 v1.1 (Illumina) scl21922.11.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980736 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21922.11.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980736 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1213549 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213549 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline