Search
Search

Cacnb1| Gene Symbol: Cacnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159319.2 | NP_001152791.1 | 8 - 9 | 933 | 57 | |
| NM_001159320.2 | NP_001152792.1 | 7 - 8 | 795 | 57 | ||
| NM_001282977.1 | NP_001269906.1 | 8 - 9 | 1068 | 57 | ||
| NM_001282978.1 | NP_001269907.1 | 7 - 8 | 909 | 57 | ||
| NM_031173.4 | NP_112450.1 | 8 - 9 | 1068 | 57 | ||
| NM_145121.3 | NP_660099.2 | 8 - 9 | 933 | 57 | ||
| GenBank mRNA | AB685743.1 | - | 8 - 9 | 1052 | 57 | |
| AF322905.1 | - | 8 - 9 | 1072 | 57 | ||
| AI466668.1 | - | 3 - 4 | 299 | 57 | ||
| AK036404.1 | - | 7 - 8 | 796 | 57 | ||
| AK155193.1 | - | 8 - 9 | 1069 | 57 | ||
| AY094172.1 | - | 8 - 9 | 731 | 57 | ||
| AY094173.1 | - | 8 - 9 | 731 | 57 | ||
| BC077713.1 | - | 7 - 8 | 905 | 30288369 | 57 | |
| CA316737.1 | - | 7 - 8 | 627 | 57 | ||
Set Membership: |
|