Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00519321_m1
          See other LYPD3 GE Assays ›
          Gene Symbol:
          Lypd3
          Gene Name:
          Ly6/Plaur domain containing 3
          Gene Aliases:
          2310061G07Rik, C4.4a
          Chromosome Location:
          Chr.7: 24636570 - 24641118 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00519321_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lypd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133743.1 NP_598504.1 4 - 5 569 67
          GenBank mRNA AK009998.1 - 4 - 5 589 67
          BC003749.1 - 4 - 5 571 67
          BC016549.1 - 4 - 5 569 3585545 67
          BC094282.1 - 4 - 5 606 5029753 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000391.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Mouse Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lypd3

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          cell-matrix adhesion
          laminin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005875 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005874 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005873 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150876 BC003749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150876 BC016549.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150876 BC094282.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150876 AK009998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150876 NM_133743.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150876 BC003749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150876 BC016549.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150876 BC094282.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150876 AK009998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150876 NM_133743.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 243246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110307 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160535 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150876 BC003749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150876 BC016549.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150876 BC094282.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150876 AK009998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150876 NM_133743.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150876 BC003749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150876 BC016549.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150876 BC094282.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150876 AK009998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150876 NM_133743.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150876 BC003749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150876 BC016549.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150876 BC094282.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150876 AK009998.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150876 NM_133743.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150876 BC003749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150876 BC016549.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150876 BC094282.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150876 AK009998.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P150876 NM_133743.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005898 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160535 - -
          Whole Mouse Genome G4122A A_51_P150876 BC003749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150876 BC016549.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150876 BC094282.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150876 AK009998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150876 NM_133743.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150876 BC003749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150876 BC016549.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150876 BC094282.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150876 AK009998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150876 NM_133743.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133615 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114661 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112736 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160535 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05888258 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05938104 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005873 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005875 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06439779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06158934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06432558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05999338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06116316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06134249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06280520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05888253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06376907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06280504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05888258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06376900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25005908 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550984 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550983 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550982 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550985 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550981 - -
          Affymetrix HT MG-430 PM Array Plate 1417942_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550980 - -
          Mouse Genome 430 2.0 Array 1417942_at - -
          Mouse Genome 430 2.0 Array 1417942_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72434_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72434_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700002893.mm.1 TC0700000391.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002893.mm.1 NM_133743.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002893.mm.1 NM_133743.1 Refseq
          Mouse Expression Set 430 1417942_at - -
          Mouse Expression Set 430 1417942_PM_at - -
          Murine Genome U74v2 Set 169732_PM_i_at - -
          Murine Genome U74v2 Set 169732_i_at - -
          Mouse Genome 430A 2.0 Array 1417942_PM_at - -
          Mouse Genome 430A 2.0 Array 1417942_at - -
          Affymetrix HT Mouse Genome 430A Array 1417942_at - -
          Affymetrix HT Mouse Genome 430A Array 1417942_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 883261 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4120288 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004120288 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248916 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248916 - -
          mouse-6 v1.1 (Illumina) scl32952.5_582-S - -
          Illumina Sentrix mouse-6.v1 bead array 4120288 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32952.5_582-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4120288 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248916 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248916 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline