Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00519903_m1
          See other ARL6IP4 GE Assays ›
          Gene Symbol:
          Arl6ip4
          Gene Name:
          ADP-ribosylation factor-like 6 interacting protein 4
          Gene Aliases:
          AA408210, AA408365, Aip-4, Srp25
          Chromosome Location:
          Chr.5: 124116108 - 124118195 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00519903_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arl6ip4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144509.2 NP_653092.1 3 - 4 904 110
          GenBank mRNA AB035383.1 - 3 - 4 904 110
          AK133934.1 - 3 - 4 720 110
          BC063748.1 - 3 - 4 871 6819350 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001437.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arl6ip4

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          RNA splicing
          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24059367 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24059365 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24059366 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24059364 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24059369 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137157 BC063748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137157 NM_144509.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137157 AB035383.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P454666 AF133911.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P454666 AF133911.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965035 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685839 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138517 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125363 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626927 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 332559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300239 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458367 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 97412 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2904 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 56544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121311 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155536 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109324 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133938 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P454666 AF133911.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P454666 AF133911.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137157 BC063748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137157 NM_144509.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137157 AB035383.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24059378 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155536 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05729417 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05729491 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05729356 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109324 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133938 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01210559 - -
          Whole Mouse Genome G4122A A_52_P533065 BC063748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533065 NM_144509.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533065 AB035383.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533065 AK133934.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P454666 AF133911.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P454666 AF133911.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137157 BC063748.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137157 NM_144509.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137157 AB035383.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109324 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133938 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155536 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01210559 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04531912 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04711904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04219058 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04711913 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04711915 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04711907 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24059367 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24059366 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24059364 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24059365 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24059369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24059332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24059383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04711915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24059382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04711904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04219058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24059373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04531901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24059372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24059376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24059368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24059378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24059367 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10525724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525721 - -
          Affymetrix Mouse Gene 1.1 ST Array 10525718 - -
          Mouse Genome 430 2.0 Array 1424285_PM_s_at - -
          Mouse Genome 430 2.0 Array 1424285_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 65105_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 65105_at - -
          Affymetrix Mouse Clariom S Array PSR0500012887.mm.1 TC0500001437.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012887.mm.1 NM_144509.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500012887.mm.1 NM_144509.2 Refseq
          Mouse Expression Set 430 1424285_s_at - -
          Mouse Expression Set 430 1424285_PM_s_at - -
          Murine Genome U74v2 Set 95136_PM_at - -
          Murine Genome U74v2 Set 95136_at - -
          Mouse Genome 430A 2.0 Array 1424285_s_at - -
          Mouse Genome 430A 2.0 Array 1424285_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 813866 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5690193 - -
          Illumina mouse-6 v1.0 expression beadchip 5690193 - -
          Illumina mouse-6 v1.0 expression beadchip 3800021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003800021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005690193 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2760116 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2740176 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2760116 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2740176 - -
          mouse-6 v1.1 (Illumina) scl27224.3.7_29-S - -
          mouse-6 v1.1 (Illumina) scl0004030.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 5690193 - -
          Illumina Sentrix mouse-6.v1 bead array 3800021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0004030.1_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27224.3.7_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690193 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3800021 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3108440 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3034691 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2760116 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2740176 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2760116 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3108440 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2740176 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3034691 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0004030.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27224.3.7_29-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline