| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369258 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369299 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369303 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369256 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369297 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369255 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369254 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369298 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369305 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1980408 |
BC030919.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1980408 |
BC042657.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1980408 |
AK045714.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1980401 |
AK049998.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1980401 |
AK134135.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1980401 |
BC030919.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1980401 |
AK045714.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1980401 |
AY453793.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
288370 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
501252 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
540232 |
AK017493.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
540232 |
AK028696.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
540232 |
AK076556.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
540232 |
BC023321.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
540232 |
BC035524.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
540232 |
NM_026484.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
540232 |
XM_001473618.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
540232 |
XM_001473645.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
132709 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
224971 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
687510 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
698186 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
800163 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
456079 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
585313 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
108033 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P121550 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P142905 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1980408 |
BC030919.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1980408 |
BC042657.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1980408 |
AK045714.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1980401 |
AK049998.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1980401 |
AK134135.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1980401 |
BC030919.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1980401 |
AK045714.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1980401 |
AY453793.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P263774 |
AK049998.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P263774 |
AK134135.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P263774 |
BC030919.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P263774 |
BC042657.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P263774 |
AK045714.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369279 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369304 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369302 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369307 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369291 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369312 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369320 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369324 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369315 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369318 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369300 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369321 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369293 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369322 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369292 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369309 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369285 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369326 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369306 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369294 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369295 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369314 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369290 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369280 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369289 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369298 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369297 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369327 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369277 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369283 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30369301 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P121550 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P142905 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P263774 |
AK049998.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263774 |
AK134135.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263774 |
BC030919.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263774 |
BC042657.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263774 |
AK045714.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115804 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P142905 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P121550 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11972358 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11972356 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11972357 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11951830 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369297 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369298 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369299 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369305 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369254 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369303 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369258 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369256 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30369255 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369263 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369275 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369205 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369248 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369265 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369317 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369171 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369142 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369180 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369294 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369176 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369206 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369141 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369304 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369260 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369279 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369312 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369338 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369289 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369200 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369168 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369196 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369137 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369175 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369285 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369340 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369266 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369156 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369203 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369245 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369326 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369140 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369291 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369298 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369235 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369239 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369198 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369146 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369314 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369230 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369227 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369183 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369274 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369213 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369221 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369215 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369241 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369144 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369190 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369293 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369231 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369280 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369256 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369251 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369254 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369191 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369259 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369330 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369163 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369316 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369277 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369253 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369331 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369136 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369173 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369320 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369295 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369134 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369178 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369219 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369302 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369226 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369216 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369133 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369306 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369229 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369272 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369188 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369193 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369204 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369283 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369328 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369268 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369310 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369267 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369333 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369300 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369238 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369224 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369184 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369234 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369166 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369212 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369194 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369160 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369318 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30369243 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12061770 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369298 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369229 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369274 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369304 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11972357 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369248 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369294 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369230 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369293 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12440667 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369260 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369256 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369268 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369232 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369324 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369330 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12011343 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369241 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369231 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369237 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369326 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12228912 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369306 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369251 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369335 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369322 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369246 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369291 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369222 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369239 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369302 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12467414 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369259 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369234 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12061772 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369263 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369272 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369243 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369266 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369307 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369254 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369323 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12207866 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369316 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369285 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369275 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12427908 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369331 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369253 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369283 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369290 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12013281 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369327 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369279 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369265 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369277 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369312 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369295 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369280 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369320 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12401772 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369224 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369333 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12207867 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12011332 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369245 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30369309 |
- |
- |