Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00520567_m1
          See other KRI1 GE Assays ›
          Gene Symbol:
          Kri1
          Gene Name:
          KRI1 homolog (S. cerevisiae)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 21273457 - 21287969 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00520567_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kri1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145416.3 NP_663391.2 4 - 5 418 79
          GenBank mRNA AK051997.1 - 4 - 5 419 79
          BC021438.1 - 4 - 5 411 5039543 79
          CJ069606.1 - 4 - 5 420 79
          CO426049.1 - 1 - 2 61 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001866.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kri1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346368 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346367 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346369 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346371 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346365 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346372 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346370 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127439 BC030861.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127439 BC069851.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127439 AK081605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127439 AK087258.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127439 AK154130.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755254 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 273317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853506 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919457 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 97986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160583 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150466 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162938 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127439 BC030861.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127439 BC069851.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127439 AK081605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127439 AK087258.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127439 AK154130.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346358 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01881858 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01881846 - -
          Whole Mouse Genome G4122A A_51_P112644 BC021438.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116702 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116505 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01881858 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07470084 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07470113 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346372 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346365 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346371 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346367 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346369 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346370 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07524244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07929280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07479107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07698388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07697599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07470113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07791029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07470104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346327 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10591502 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591504 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591500 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583583 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591501 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591503 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591505 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591508 - -
          Affymetrix HT MG-430 PM Array Plate 1424788_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1442872_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10583573 - -
          Affymetrix Mouse Gene 1.1 ST Array 10591497 - -
          Mouse Genome 430 2.0 Array 1420135_at - -
          Mouse Genome 430 2.0 Array 1424788_at - -
          Mouse Genome 430 2.0 Array 1449743_PM_a_at - -
          Mouse Genome 430 2.0 Array 1424788_PM_at - -
          Mouse Genome 430 2.0 Array 1449743_a_at - -
          Mouse Genome 430 2.0 Array 1442872_at - -
          Mouse Genome 430 2.0 Array 1442872_PM_at - -
          Mouse Genome 430 2.0 Array 1420135_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 215194_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 215194_at - -
          Affymetrix Mouse Clariom S Array PSR0900015703.mm.1 NM_145416.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015703.mm.1 NM_145416.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015703.mm.1 TC0900001866.mm.1 MTA TC
          Mouse Expression Set 430 1424788_PM_at - -
          Mouse Expression Set 430 1424788_at - -
          Mouse Expression Set 430 1442872_at - -
          Mouse Expression Set 430 1442872_PM_at - -
          Mouse Genome 430A 2.0 Array 1424788_PM_at - -
          Mouse Genome 430A 2.0 Array 1424788_at - -
          Affymetrix HT Mouse Genome 430A Array 1424788_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424788_at - -
          Affymetrix HT Mouse Genome 430B Array 1442872_at - -
          Affymetrix HT Mouse Genome 430B Array 1442872_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1780195 - -
          MouseWG-6 v2 Expression BeadChip 106520537 - -
          MouseWG-6 v2 Expression BeadChip 6940170 - -
          Illumina mouse-6 v1.0 expression beadchip 106520537 - -
          Illumina mouse-6 v1.0 expression beadchip 6940170 - -
          Illumina mouse-6 v1.0 expression beadchip 1780195 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780195 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940170 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2644354 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243563 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2644354 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2486702 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243563 - -
          mouse-6 v1.1 (Illumina) scl0003523.1_1-S - -
          mouse-6 v1.1 (Illumina) scl37227.2.1_30-S - -
          mouse-6 v1.1 (Illumina) scl36147.14.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 106520537 - -
          Illumina Sentrix mouse-6.v1 bead array 1780195 - -
          Illumina Sentrix mouse-6.v1 bead array 6940170 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36147.14.1_30-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003523.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106520537 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780195 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940170 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2486702 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605740 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline